Skip to Content
Merck
All Photos(3)

Documents

N9125

Sigma-Aldrich

N-(1-Naphthyl)ethylenediamine dihydrochloride

≥98%

Synonym(s):

2-(1-Naphthylamino)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
Beilstein:
3707471
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

194-198 °C (dec.) (lit.)

SMILES string

Cl.Cl.NCCNc1cccc2ccccc12

InChI

1S/C12H14N2.2ClH/c13-8-9-14-12-7-3-5-10-4-1-2-6-11(10)12;;/h1-7,14H,8-9,13H2;2*1H

InChI key

MZNYWPRCVDMOJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(1-Naphthyl)ethylenediamine dihydrochloride can be used as a reactant to synthesize:      
  • N


  • -(1-naphthyl)ethylenediamine-substituted cylotriphosphazene derivatives as potential antimicrobial agents.    
  • Fluorescent chemosensor compound with a naphthyl group as fluorophore and cyclen as metal ion chelator for the detection of Zn(II) in an aqueous solution.    
  • (N-1-Naphthyl-ethylenediamine)- dichloroplatinum(II) fluorescence complex (λmax = 405 nm).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Morais de Paiva Daibert et al.
PloS one, 15(11), e0241861-e0241861 (2020-11-07)
Macrophages are classified upon activation as classical activated M1 and M2 anti-inflammatory regulatory populations. This macrophage polarization is well characterized in humans and mice, but M1/M2 profile in cattle has been far less explored. Bos primigenius taurus (taurine) and Bos
C E Davis et al.
Journal - Association of Official Analytical Chemists, 68(3), 485-488 (1985-05-01)
Current methods used for determining residual nitrite concentration in foods involve forming an azo dye that is measured spectrophotometrically. Conventional procedures do not specify control of pH for the final colored solution. Because many indicator dyes are pH-dependent, absorbance of
Improved dye procedure for determining urea concentration by using o-phthalaldehyde and naphthylethylenediamine.
N T Lequang et al.
Clinical chemistry, 33(1), 192-192 (1987-01-01)
P K Laikind et al.
Analytical biochemistry, 156(1), 81-90 (1986-07-01)
The Bratton-Marshall reaction can be used to identify patients with adenylosuccinate lyase deficiency. These patients excrete in their urine the dephosphorylated derivative of the de novo purine synthesis intermediate 5'-phosphoribosyl-4-(N-succinylcarboxamide)-5-aminoimidazole (SAICAR). The test described here depends on a coupling reaction
J A Cook et al.
Analytical biochemistry, 238(2), 150-158 (1996-07-01)
S-nitrosothiols have been shown to affect a number of physiological functions. Several techniques have been used to detect these species in biological systems, primarily by methods utilizing chemiluminescence. Since the apparatus required for measurement of chemiluminescence are not readily available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service