Skip to Content
Merck
All Photos(1)

Key Documents

T6700

Sigma-Aldrich

TRAM-34

≥98% (HPLC), solid

Synonym(s):

1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: 2 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4cccn4

InChI

1S/C22H17ClN2/c23-21-15-8-7-14-20(21)22(25-17-9-16-24-25,18-10-3-1-4-11-18)19-12-5-2-6-13-19/h1-17H

InChI key

KBFUQFVFYYBHBT-UHFFFAOYSA-N

Related Categories

Application

TRAM-34 has been used to study its effect on the prevention of vascular restenosis after balloon angioplasty.

Biochem/physiol Actions

TRAM-34 exhibits 100-fold selectivity (Kd = 20 nM) for IKCa1 channels over other K+ channels (Charybdotoxin, Kd = 5 nM, Clotrimazole, Kd = 70 nM; Nitrendipine Kd = 900 nM). Furthermore, TRAM-34 is known to suppress the activation of human T lymphocytes,.
TRAM-34 is a potent inhibitor of the intermediate-conductance Ca2+-activated K+ channel.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

TRAM-34 is soluble in DMSO at 2 mg/ml. However, it is insoluble in water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friederike A Steudel et al.
Molecular oncology, 11(9), 1172-1188 (2017-05-31)
Oncogenic signalling via Ca
C Kondo et al.
Experimental physiology, 103(8), 1101-1122 (2018-05-24)
What is the central question of this study? What are the main [Ca2+ ]i signalling pathways activated by ATP in human synovial fibroblasts? What is the main finding and its importance? In human synovial fibroblasts ATP acts through a linked
Nicole Glaser et al.
Pediatric diabetes, 18(5), 356-366 (2016-05-14)
Diabetic ketoacidosis (DKA) causes brain injuries in children ranging from subtle to life-threatening. Previous studies suggest that DKA-related brain injury may involve both stimulation of Na-K-Cl cotransport and microglial activation. Other studies implicate the Na-K-Cl cotransporter and the Ca-activated K
Génesis Vega et al.
JCI insight, 5(16) (2020-08-21)
Airway mucociliary clearance (MCC) is the main mechanism of lung defense keeping airways free of infection and mucus obstruction. Airway surface liquid volume, ciliary beating, and mucus are central for proper MCC and critically regulated by sodium absorption and anion
H Wulff et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(14), 8151-8156 (2000-07-08)
The antimycotic clotrimazole, a potent inhibitor of the intermediate-conductance calcium-activated K(+) channel, IKCa1, is in clinical trials for the treatment of sickle cell disease and diarrhea and is effective in ameliorating the symptoms of rheumatoid arthritis. However, inhibition of cytochrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service