Skip to Content
Merck
All Photos(1)

Key Documents

U7752

Sigma-Aldrich

Urease from Canavalia ensiformis (Jack bean)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

biological source

Canavalia ensiformis

Quality Level

form

solid

mol wt

hexamer 545 kDa
trimer ~272 kDa

contains

~20% dithiothreitol

suitability

suitable for marker for electrophoresis (non-denaturing PAGE)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Subunit molecular weight: ~90,770
Composed of six subunits with total molecular weight: ~544,620
Urease is a multi-subunit nickel-dependent metalloenzyme. It is found abundantly in fungi, yeast, plants, and bacteria. It belongs to the superfamily of amidohydrolases and phosphotriesterases. Urease is also found in leguminous seeds.

Application

Urease from Canavalia ensiformis (Jack bean) has been used:
  • as a component of the molecular mass standard in electrophoresis
  • as a component of the standard in size-exclusion chromatography
  • as a reference in native-polyacrylamide gel electrophoresis (PAGE)

Biochem/physiol Actions

Urease is an essential enzyme of the nitrogen cycle. It plays a role in degrading urea into ammonia and carbamate. Urease provides a source of nitrogen for growth to various organisms. It also plays a major role in seed germination and seed chemical defense.

Other Notes

Contains approx. 1 mg protein

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Ajmal Rana et al.
PloS one, 16(10), e0258568-e0258568 (2021-10-15)
Urea is the most popular and widely used nitrogenous fertilizer. High soil urease activity rapidly hydrolyses applied urea to ammonia which contributes to soil nitrogen (N) losses and reduces N use efficiency of crop plants. The ammonia losses can be
Anuradha Balasubramanian et al.
Journal of molecular biology, 400(3), 274-283 (2010-05-18)
Urease, a nickel-dependent metalloenzyme, is synthesized by plants, some bacteria, and fungi. It catalyzes the hydrolysis of urea into ammonia and carbon dioxide. Although the amino acid sequences of plant and bacterial ureases are closely related, some biological activities differ
Jonathan Parkinson et al.
Nature communications, 14(1), 454-454 (2023-01-29)
High-affinity antibodies are often identified through directed evolution, which may require many iterations of mutagenesis and selection to find an optimal candidate. Deep learning techniques hold the potential to accelerate this process but the existing methods cannot provide the confidence
Karine Kappaun et al.
Journal of advanced research, 13, 3-17 (2018-08-11)
Urease (urea amidohydrolase, EC 3.5.1.5) is a nickel-containing enzyme produced by plants, fungi, and bacteria that catalyzes the hydrolysis of urea into ammonia and carbamate. Urease is of historical importance in Biochemistry as it was the first enzyme ever to
Enhanced catalytic properties of novel (alphagamma) 2 heterohexameric Rhodobacter capsulatus xanthine dehydrogenase by separate expression of the redox domains in Escherichia coli
Wang C, et al.
Biochemical Engineering Journal, 119, 1-8 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service