Skip to Content
Merck
All Photos(1)

Key Documents

M7257

Sigma-Aldrich

α-Mannosidase from Canavalia ensiformis (Jack bean)

ammonium sulfate suspension, ≥15 units/mg protein (biuret)

Synonym(s):

α-D-Mannoside mannohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Canavalia ensiformis

Quality Level

form

ammonium sulfate suspension

specific activity

≥15 units/mg protein (biuret)

foreign activity

β-galactosidase, β-N-acetylglucosaminidase, α-galactosidase and α-L-fucosidase <0.05%

storage temp.

2-8°C

SMILES string

[Zn+2]

InChI

1S/Zn/q+2

InChI key

PTFCDOFLOPIGGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α−Mannosidase from Canavalia ensiformis is a tretamer composed of two subunits that each contain two components at 44 and 66 kDa.
α-Mannosidase is an exo-glycosidase and an inc metalloenzyme.
Lysosomal α-mannosidases are class II mannosidases that belong to glycoside hydrolase family 38 and play an important role in the degradation of asparagine-linked carbohydrates of glycoproteins.

Application

α-Mannosidase can be used to liberate mannose from a variety of synthetic and natural α-mannosides. It has also been used in a study to investigate the causes of neurodegeneration in mucolipidosis II ′knock-in′ mice.
α-Mannosidase from Canavalia ensiformis (Jack bean) has been used:
  • for the digestion of neutral glycans.
  • to remove O-glycans from recombinant β-glucosidase
  • for the inhibition of renal fibroblasts proliferation

Liberates mannose from a variety of synthetic and natural α-mannosides.

Biochem/physiol Actions

α−Mannosidase has been shown to inhibit the proliferation of B-lymphocytes.
α-Mannosidase cleaves near α1−2-, α1−3- and α1−6-linked mannose residues.
α-Mannosidase is an acid hydrolase which is located in plant vacuoles and is thought to be involved with the turnover of N-linked glycoproteins.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl α-D-mannoside to p-nitrophenol and D-mannose per min at pH 4.5 at 25°C.

Physical form

Suspension in 3.0 M (NH4)2SO4 and 0.1 mM zinc acetate, pH 7.5

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reto Buser et al.
The Journal of biological chemistry, 285(14), 10715-10723 (2010-01-12)
Coprinopsis cinerea is a model organism for fruiting body development in homobasidiomycetes. Here, we focused on N-linked oligosaccharides (NLO) of cell wall proteins in the hyphae of two developmental stages, vegetative mycelium and fruiting body. High mannose-type glycans were the
Jack bean alpha-mannosidase: Amino acid sequencing and N-glycosylation analysis of a valuable glycomics tool.
Kumar G, et al.
Glycobiology, 24(3) (2014)
Engineering a highly active thermophilic beta-glucosidase to enhance its pH stability and saccharification performance
Xia W, et al.
Biotechnology for Biofuels, 9(1), 147-147 (2016)
A Family of Dual-Activity Glycosyltransferase-Phosphorylases Mediates Mannogen Turnover and Virulence in Leishmania Parasites
Sernee MF, et al.
Cell host & microbe, 26, 385-399 (2019)
Collectin-11 promotes the development of renal tubulointerstitial fibrosis
Wu W, et al.
Journal of the American Society of Nephrology, 29, 168-181 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service