Skip to Content
Merck
All Photos(4)

Documents

U1500

Sigma-Aldrich

Urease from Canavalia ensiformis (Jack bean)

Type III, powder, 15,000-50,000 units/g solid

Synonym(s):

Jack bean urease, Urea amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

biological source

Canavalia ensiformis

type

Type III

form

powder

specific activity

15,000-50,000 units/g solid

mol wt

~544620 Da

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Subunit molecular weight: ~90,770
Composed of six subunits with total molecular weight: ~544,620
Urease is a cytosolic enzyme that catalyzes the hydrolysis of urea to ammonia and carbon dioxide, with initial conversion to carbamate, and the carbamate breaks down further, on hydrolysis, into ammonia and to carbonic acid / carbonate. Urease is a metalloenzyme that contains nickel as a co-factor. Urease occurs naturally in plants, fungi, and certain bacteria. James B. Sumner first isolated urease from jack bean in 1926.

Biochem/physiol Actions

Urease from Canavalia ensiformis functions as an insecticide by the release of a toxic small peptide cleaved by proteases found in the gut of insects.

Unit Definition

One micromolar unit will liberate 1.0 μmole of NH3 from urea per min at pH 7.0 at 25 °C. It is equivalent to 1.0 I.U. or 0.054 Sumner unit (1.0 mg ammonia nitrogen in 5 minutes at pH 7.0 at 20 °C)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han-Ho Kim et al.
Journal of microbiology and biotechnology, 31(12), 1684-1691 (2021-12-29)
In this study, the physicochemical changes related to fishy smell were determined by storing high hydrostatic pressure (HHP)-treated mackerel (Scomber japonicus) meat in a refrigerator for 20 days. The inhibition of crude urease activity from Vibrio parahaemolyticus using HHP treatment
Xiang-Zhen Yan et al.
Macromolecular bioscience, 14(7), 1004-1014 (2014-04-02)
The aim of this study is to optimize enzymatic control over gelation of chitosan-based hydrogels for the delivery of periodontal ligament cells (PDLCs). The results reveal that the gelation time, strength, and degradation rate of the chitosan hydrogels can be
Fernanda Stanisçuaski et al.
Journal of insect physiology, 55(3), 255-263 (2009-01-06)
Ureases are metalloenzymes that are widespread among plants, fungi and bacteria. Urease isoforms (jack bean urease-JBU and canatoxin) from Canavalia ensiformis seeds are toxic to insects and fungi, suggesting a role in plant defense. The entomotoxic effect is due to
Sun-Il Kim et al.
Microorganisms, 10(1) (2022-01-22)
Ammonia from livestock manure reacts with chemical components discharged from various emission sources to produce airborne particulate matter. This study aimed to investigate a novel effective microbial agent to suppress ammonia gas emitted from manure. Both isolated L12I and 12III
Arlette Yumba-Mpanga et al.
Current medicinal chemistry, 26(1), 216-231 (2017-10-11)
In regard to urogenital tract cancer studies, an estimated 340,650 new cases and 58,360 deaths from genital system cancer and about 141,140 new cases and 29330 deaths from urinary system were projected to occur in the United States in 2012.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service