The type designations for urease products are not indications of particular isoforms. They are internal codes that refer to the chronological order of the products' market entry and manufacturing processes. The manufacturing processes are proprietary. However, the major differences between the urease products are their specifications for specific activity. Please access the 'Specification Sheet' and 'Sample Certificate of Analysis' for each individual product via its corresponding website.
U4002
Urease from Canavalia ensiformis (Jack bean)
Type IX, powder, 50,000-100,000 units/g solid
Synonym(s):
Jack bean urease, Urea amidohydrolase
About This Item
Recommended Products
type
Type IX
Quality Level
form
powder
specific activity
50,000-100,000 units/g solid
mol wt
~544620 Da
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Composed of six subunits with total molecular weight: ~544,620
Application
Biochem/physiol Actions
Unit Definition
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Good morning, I would like to know what the difference is between the different types of urease, since some are type III, type IX, type C-III.
1 answer-
Helpful?
-
-
Hi there I was wondering what Type IX means and how many grams are there in the 20 kU one?
1 answer-
The 'type' is a historical designation used to differentiate various Urease products. Each lot has a specific activity of 50,000 - 100,000 units per gram solid. Therefore, the mass of the 20 kU option can vary between 0.2 g and 0.4 g. The lot-specific certificate of analysis will provide the specific activity.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service