Skip to Content
Merck
All Photos(2)

Key Documents

U6756

Sigma-Aldrich

U-73122 hydrate

powder

Synonym(s):

1-[6-[((17β)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H40N2O3 · xH2O
CAS Number:
Molecular Weight:
464.64 (anhydrous basis)
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97.5% (HPLC)

form

powder

color

off-white

solubility

ethanol: ~0.7 mg/mL
DMSO: ~0.9 mg/mL
chloroform: ~10 mg/mL
DMSO: ≤2.6 mg/mL (Achieved with heating.)
ethanol: ≤5 mg/mL (Achieved with heating.)
H2O: insoluble

storage temp.

room temp

SMILES string

O.COc1ccc2C3CC[C@]4(C)[C@H](CCC4C3CCc2c1)NCCCCCCN5C(=O)C=CC5=O

InChI

1S/C29H40N2O3.H2O/c1-29-16-15-23-22-10-8-21(34-2)19-20(22)7-9-24(23)25(29)11-12-26(29)30-17-5-3-4-6-18-31-27(32)13-14-28(31)33;/h8,10,13-14,19,23-26,30H,3-7,9,11-12,15-18H2,1-2H3;1H2/t23-,24-,25+,26+,29+;/m1./s1

InChI key

WCOYJDRCHHGMRK-BDEPKWELSA-N

Looking for similar products? Visit Product Comparison Guide

Application

U-73122 hydrate has been used for studying the role of phospholipase C (PLC) signaling in modulating epithelial Na+ channel (ENaC) function in gerbil Reissner′s membranes (RMs). U-73122 has also been used to study local calcium responses in human umbilical vein endothelial cells (HUVECs).

Biochem/physiol Actions

Inhibits the hydrolysis of PPI to IP3, which leads to a decrease in cytosolic free calcium. Inhibits the coupling of G protein-phospholipase C activation, while remaining unaffected by production of cAMP.

Features and Benefits

This compound is featured on the Phospholipase C page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

U-73122 hydrate is soluble in ethanol (approx. 0.7 mg/ml or ≤ 5 mg/ml with heating), chloroform (approx. 10 mg/ml) and DMSO (approx. 0.9 mg/ml or ≤ 2.6 mg/ml with heating). U-73122 hydrate is insoluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Sung Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(37), 15079-15084 (2013-08-28)
Recruitment of release-competent vesicles during sustained synaptic activity is one of the major factors governing short-term plasticity. During bursts of synaptic activity, vesicles are recruited to a fast-releasing pool from a reluctant vesicle pool through an actin-dependent mechanism. We now
D I Yule et al.
The Journal of biological chemistry, 267(20), 13830-13835 (1992-07-15)
Stimulation of rat pancreatic acinar cells with low concentrations of phosphatidylinositol (PI)-linked secretagogues induces [Ca2+]i oscillations, without measurable changes in the formation of inositol 1,4,5-trisphosphate. Therefore, we tested U73122 a new phospholipase C inhibitor to determine if PI turnover is
Wagner Shin Nishitani et al.
PloS one, 6(10), e26181-e26181 (2011-11-05)
A new device was designed to generate a localized mechanical vibration of flexible gels where human umbilical vein endothelial cells (HUVECs) were cultured to mechanically stimulate these cells at subcellular locations. A Fluorescence Resonance Energy Transfer (FRET)-based calcium biosensor (an
Wei Fu et al.
Frontiers in endocrinology, 10, 930-930 (2020-02-23)
Plasma levels of PCSK9 are significantly higher in postmenopausal women. Pharmacologically increased estrogen levels have been shown to lower PCSK9 and LDL-C levels in animals and humans. The action of estrogen suggests that it has the ability to prevent PCSK9-mediated
R C Smallridge et al.
Endocrinology, 131(4), 1883-1888 (1992-10-01)
TRH increases cytosolic-free calcium ([Ca2+]i) by activating phospholipase C(PL-C), which induces phosphoinositol hydrolysis, leading to Ca2+ mobilization from inositol trisphosphate (IP3) sensitive stores, and by increasing Ca2+ influx. Increases in [Ca2+]i stimulate PRL secretion. We investigated the effects of U-73122

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service