Skip to Content
Merck
All Photos(3)

Key Documents

C1530

Sigma-Aldrich

Cyclopiazonic acid

from Penicillium cyclopium, ≥98% (HPLC), powder, sarcoplasmic reticulum Ca²⁺-pump inhibitor

Synonym(s):

CPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2O3
CAS Number:
Molecular Weight:
336.38
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Product Name

Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: soluble
chloroform: soluble

storage temp.

−20°C

SMILES string

CN1C(=O)C(=C(\C)O)\C(=O)C2C(Cc3cccc4[nH]cc2c34)C1(C)C

InChI

1S/C20H22N2O3/c1-10(23)15-18(24)17-12-9-21-14-7-5-6-11(16(12)14)8-13(17)20(2,3)22(4)19(15)25/h5-7,9,13,17,21,23H,8H2,1-4H3/b15-10-

InChI key

WJCPGKDPOYBOOJ-GDNBJRDFSA-N

General description

Cyclopiazonic acid (CPA), a tetramic acid is synthesized by various Aspergillus and Penicillium species.

Application

Cyclopiazonic acid from Penicillium cyclopium has been used:
  • as a standard in aflatoxin detection and quantification by thin layer chromatography (TLC)
  • as a component in Ca2+-free DMEM/10% fetal bovine serum (FBS)
  • as a reversible inhibitor of the sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) to examine the role of luminal calcium in endoplasmic recticulum (ER)-to-Golgi trafficking
  • as an endoplasmic reticulum Ca2+-ATPase blocker, to treat neurons and also to study the role of the endoplasmic reticulum (ER) in releasing and sustaining Ca2+ during Ca2+-induced Ca2+ release (CICR) to inhibit SERCA

Biochem/physiol Actions

Inhibits the sarcoplasmic reticulum Ca2+-pump.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakiko Ujita et al.
Cerebral cortex (New York, N.Y. : 1991), 27(2), 1602-1614 (2016-01-24)
Astrocytes in various brain regions exhibit spontaneous intracellular calcium elevations both in vitro and in vivo; however, neither the temporal pattern underlying this activity nor its function has been fully evaluated. Here, we utilized a long-term optical imaging technique to
G W J Wright et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 24(5), e221-e234 (2012-02-02)
Interstitial cells of Cajal (ICC) associated with the myenteric plexus of the small intestine express maxi chloride channels. Our aim was to investigate whether or not these channels would be activated by increases in intracellular Ca(2+) , as that would
Ca2+-Induced Ca2+ Release in Aplysia Bag Cell Neurons Requires Interaction Between Mitochondrial and Endoplasmic Reticulum Stores
Geiger JE, et al.
Journal of Neurophysiology (2008)
Toxigenic Aspergillus flavus and other fungi of public health concern in food and organic matter in southwest Nigeria
Fapohunda SO, et al.
Mycology, 3, 210-219 (2012)
N Demaurex et al.
The Journal of biological chemistry, 267(4), 2318-2324 (1992-02-05)
The filling state of intracellular Ca2+ stores has been proposed to regulate Ca2+ influx across the plasma membrane in a variety of tissues. To test this hypothesis, we have used three structurally unrelated inhibitors of the Ca(2+)-ATPase of intracellular Ca2+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service