Skip to Content
Merck
All Photos(1)

Documents

662035

Sigma-Aldrich

U-73122

≥98% (HPLC), solid, agonist-induced phospholipase C activation inhibitor, Calbiochem®

Synonym(s):

U-73122, 1-[6-((17β-3-Methoxyestra-1,3,5(10)-trien-17-yl)amino)hexyl]-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H40N2O3
CAS Number:
Molecular Weight:
464.64
UNSPSC Code:
41121800
NACRES:
NA.77

product name

U-73122, U-73122, CAS 112648-68-7, inhibits agonist-induced phospholipase C activation (IC50 = 1-2.1 µM) in human platelets and neutrophils.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

anhydrous DMSO: 1 mg/mL
chloroform: 3 mg/mL
anhydrous ethanol: 500 μg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C29H40N2O3/c1-29-16-15-23-22-10-8-21(34-2)19-20(22)7-9-24(23)25(29)11-12-26(29)30-17-5-3-4-6-18-31-27(32)13-14-28(31)33/h8,10,13-14,19,23-26,30H,3-7,9,11-12,15-18H2,1-2H3/t23-,24-,25+,26+,29+/m1/s1

InChI key

LUFAORPFSVMJIW-ZRJUGLEFSA-N

General description

A potent, selective, and cell-permeable inhibitor C-type phosphatidylinositol-specific phospholipases. Inhibits agonist-induced phospholipase C activation (IC50 = 1-2.1 µM) in human platelets and neutrophils. Inhibits ET-1 and parathyroid hormone induced Ca2+ transients (IC50 = 800 nM and 500 nM, respectively). Also inhibits agonist-induced down-regulation of muscarinic receptors in SK-N-SH neuroblastoma cells. Acts as a potent inhibitor of human polymorphonuclear neutrophil adhesion on biological surfaces (IC50 = 50 nM). Abolishes T lymphoma cell invasion into fibroblast monolayers.
A potent, selective, and cell-permeable inhibitor C-type phosphatidylinositol-specific phospholipases. Inhibits agonist-induced phospholipase C activation (IC50 = 1-2.1 µM) in human platelets and neutrophils. Inhibits ET-1 and parathyroid hormone-induced Ca2+ transients (IC50 = 800 nM and 500 nM, respectively). Also inhibits agonist-induced down-regulation of muscarinic receptors in SK-N-SH neuroblastoma cells. Acts as a potent inhibitor of human polymorphonuclear neutrophil adhesion on biological surfaces (IC50 = 50 nM). Reported to abolish T lymphoma cell invasion into fibroblast monolayers.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Agonist-induced phospholipase C activation
Product does not compete with ATP.
Reversible: no
Target IC50: 1-2.1 µM against agonist-induced phospholipase C activation in human platelets and neutrophils; 800 nM and 500 nM, respectively, against ET-1 and parathyroid hormone-induced Ca2+ transients; 50 nM against human polymorphonuclear neutrophil adhesion

Warning

Toxicity: Irritant (B)

Preparation Note

The use of anhydrous solvents is critical for the solubility of the compound; the presence of moisture in the solvent will lower the solubility of the compound.

Reconstitution

Unstable in DMSO and ethanol; reconstitute in anhydrous DMSO or ethanol just prior to use. Following reconstitution in anhydrous chloroform, aliquot into single use volumes, evaporate to dryness under a stream of nitrogen, and freeze (-20°C. Dried aliquots are stable for up to 6 months at -20°C. ); Reconstitute dried aliquots in anhydrous DMSO or anhydrous ethanol just prior to use.

Other Notes

Stam, J.C., et al. 1998. EMBO J. 17, 4066.
Smith, R.J., et al. 1996. J. Pharmacol. Exp. Ther.278, 320.
Tatrai, A., et al. 1994. Biochim. Biophys. Acta 1224, 595.
Wang, X.D., et al. 1994. Biochim. Biophys. Acta1223, 101.
Bleasdale, J.E., and Fisher, S.K. 1993. Neuroprotocols: A Companion to Methods in Neurosci. 3, 125.
Wu, H., et al. 1992. Biochemistry31, 3370.
Yule, D.I., and Williams, J.A. 1992. J. Biol. Chem.267, 13830.
Thompson, A.K., et al. 1991. J. Biol. Chem.266, 23856.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junpei Matsubayashi et al.
Journal of neurochemistry, 164(1), 29-43 (2022-12-01)
Neurons in the central nervous system (CNS) have limited capacity for axonal regeneration after trauma and neurological disorders due to an endogenous nonpermissive environment for axon regrowth in the CNS. Lateral olfactory tract usher substance (LOTUS) contributes to axonal tract
Archna Sharma et al.
Molecular neurobiology, 60(6), 3311-3328 (2023-03-01)
Abnormal calcium homeostasis, activation of protease calpain, generation of p25 and hyperactivation of cyclin-dependent kinase 5 (Cdk5) have all been implicated in the pathogenesis of neurogenerative diseases including Alzheimer's disease. We have recently shown that extracellular cold-inducible RNA-binding protein (eCIRP)
J C Norman et al.
FEBS letters, 484(3), 179-183 (2000-11-18)
Aggregation by immune complexes of receptors specific for the Fc region of IgG results in their internalisation and disposal by trafficking to lysosomes. We show here that internalisation of FcgammaRI by IFN-gamma treated U937 cells following receptor aggregation by cross-linking
Noah Steinfeld et al.
Molecular biology of the cell, 35(1), ar5-ar5 (2023-11-01)
The interaction between aggregated low-density lipoprotein (agLDL) and macrophages in arteries plays a major role in atherosclerosis. Macrophages digest agLDL and generate free cholesterol in an extracellular, acidic, hydrolytic compartment known as the lysosomal synapse. Macrophages form a tight seal
A Soledad Coria et al.
Biology of the cell, 106(1), 30-43 (2013-11-02)
Heterotrimeric GTP-binding proteins play a key role in cell trafficking regulation. Above all, specific Gβγ subunits have been shown to be a major component of a signal transduction pathway, which also involves phospholipases C (PLC), protein kinases C (PKC) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service