Skip to Content
Merck
All Photos(2)

Key Documents

G1918

Sigma-Aldrich

Gö 6983

≥97%

Synonym(s):

3-[1-[3-(Dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N4O3
CAS Number:
Molecular Weight:
442.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97%

form

powder

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

COc1ccc2n(CCCN(C)C)cc(C3=C(C(=O)NC3=O)c4c[nH]c5ccccc45)c2c1

InChI

1S/C26H26N4O3/c1-29(2)11-6-12-30-15-20(18-13-16(33-3)9-10-22(18)30)24-23(25(31)28-26(24)32)19-14-27-21-8-5-4-7-17(19)21/h4-5,7-10,13-15,27H,6,11-12H2,1-3H3,(H,28,31,32)

InChI key

LLJJDLHGZUOMQP-UHFFFAOYSA-N

Application

GO6983 has been used as a protein kinase C (PKC) inhibitor to study its effects on the nuclear import. It has also been used as a supplement in Fc medium to culture mouse embryonic stem (mES) cells on mitomycin C treated mouse embryonic fibroblasts (MEFs).

Biochem/physiol Actions

Protein kinase C (PKC) inhibitor that selectively inhibits several PKC isozymes (IC50 = 7 nM for PKCα and PKCβ; 6 nM for PKCγ; 10 nM for PKCδ; and 60 nM for PKCζ), but does not inhibit PKCμ (IC50 = 20 mM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pluripotent state transitions coordinate morphogenesis in mouse and human embryos
Shahbazi MN, et al.
Nature, 552(7684), 239-239 (2017)
Ohad Gafni et al.
Nature, 504(7479), 282-286 (2013-11-01)
Mouse embryonic stem (ES) cells are isolated from the inner cell mass of blastocysts, and can be preserved in vitro in a naive inner-cell-mass-like configuration by providing exogenous stimulation with leukaemia inhibitory factor (LIF) and small molecule inhibition of ERK1/ERK2
Li-Chun Lisa Tsai et al.
The Journal of biological chemistry, 290(36), 21845-21856 (2015-07-19)
Atypical protein kinase C (aPKC) enzymes signal on protein scaffolds, yet how they are maintained in an active conformation on scaffolds is unclear. A myristoylated peptide based on the autoinhibitory pseudosubstrate fragment of the atypical PKCζ, zeta inhibitory peptide (ZIP)
Debasree Dutta et al.
Stem cells (Dayton, Ohio), 29(4), 618-628 (2011-02-11)
The intricate molecular mechanisms that regulate ESC pluripotency are incompletely understood. Prior research indicated that activation of the Janus kinase-signal transducer and activator of transcription (STAT3) pathway or inhibition of extracellular signal-regulated kinase/glycogen synthase kinase 3 (ERK/GSK3) signaling maintains mouse
Shirley E Bradley et al.
The Journal of biological chemistry, 285(7), 4883-4895 (2009-11-18)
Expression of the VACM-1/cul5 gene in endothelial and in cancer cell lines in vitro inhibits cellular proliferation and decreases phosphorylation of MAPK. Structure-function analysis of the VACM-1 protein sequence identified consensus sites specific for phosphorylation by protein kinases A and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service