Skip to Content
Merck
All Photos(1)

Key Documents

U104

Sigma-Aldrich

UK 14,304

Synonym(s):

5-Bromo-N-(2-imidazolin-2-yl)-6-quinoxalinamine, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine, Brimonidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BrN5
CAS Number:
Molecular Weight:
292.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: <0.8 mg/mL
DMSO: >6.5 mg/mL
ethanol: <8 mg/mL (warm)
H2O: insoluble

SMILES string

Brc1c(NC2=NCCN2)ccc3nccnc13

InChI

1S/C11H10BrN5/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8/h1-4H,5-6H2,(H2,15,16,17)

InChI key

XYLJNLCSTIOKRM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

UK 14,304 has been used for the activation of extracellular signal-regulated kinase (ERK) in epicardial coronary artery culture
UK 14,304 has been used in myenteric neurons to study the inhibition of R-type calcium currents. UK 14,304 has also been used for immunofluorescence microscopy assays.

Biochem/physiol Actions

UK 14,304 is an α2-adrenoceptor agonist. UK 14,304 inhibits hormone-sensitive lipase (HSL) activity and suppresses lipogenesis in adipose tissue.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

UK 14,304 is soluble in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (<0.8 mg/mL), DMSO (>6.5 mg/ml) and ethanol (<8 mg/mL). However, it is insoluble in water.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Wang et al.
Journal of biomedical nanotechnology, 13(9), 1089-1096 (2018-02-27)
In this work, we developed a mildly cross-linked dendrimer hydrogel (mcDH) via
Kenric Rui-Pin Fan et al.
Acta ophthalmologica, 97(6), e827-e832 (2019-03-28)
To investigate the neuroprotective effect of Copolymer-1 (Cop-1) in patients with acute primary angle closure (APAC) in a randomized double-masked controlled trial. After initial medical management, APAC patients were randomized to receive either subcutaneous Cop-1 or placebo within 24 hr and
Effects of acute and chronic ethanol administration on the response of mouse adipose tissue hormone-sensitive lipase to alpha2-adrenoceptor activation by UK 14304.
Shih MF and Taberner PV
Alcohol and Alcoholism (Oxford, Oxfordshire), 36(5), 381-387 (2001)
J T Turner et al.
Molecular pharmacology, 28(5), 422-430 (1985-11-01)
We have characterized the alpha 2-adrenergic receptor in membranes from the human colonic adenocarcinoma cell line, HT29, using the recently introduced alpha 2-agonist 5-bromo-6-[2-imidazolin-2-yl-amino]quinoxaline [( 3H]UK-14,304), two other radioligands, and a series of adrenergic agonists and antagonists. We also investigated
Hung-Yu Lin et al.
Medicine, 96(47), e8868-e8868 (2018-02-01)
To evaluate the safety and efficiency in macular edema patients who concurrently received a single injection of a dexamethasone intravitreal implant (DEX, 0.7 mg) and ranibizumab (2.3 mg).A retrospective cohort study was conducted, and medical records from 2012 to 2016 were reviewed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service