Skip to Content
Merck
All Photos(1)

Key Documents

SML0281

Sigma-Aldrich

Ivabradine hydrochloride

≥98% (HPLC)

Synonym(s):

3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36N2O5 · HCl
CAS Number:
Molecular Weight:
505.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +5 to +9°, c = 1 in DMSO

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

Cl.COc1cc2CCN(CCCN(C)C[C@H]3Cc4cc(OC)c(OC)cc34)C(=O)Cc2cc1OC

InChI

1S/C27H36N2O5.ClH/c1-28(17-21-11-20-14-25(33-4)26(34-5)16-22(20)21)8-6-9-29-10-7-18-12-23(31-2)24(32-3)13-19(18)15-27(29)30;/h12-14,16,21H,6-11,15,17H2,1-5H3;1H/t21-;/m1./s1

InChI key

HLUKNZUABFFNQS-ZMBIFBSDSA-N

Gene Information

human ... HCN4(10021)

Application

Ivabradine hydrochloride has been used as a potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel (HCN)2 blocker in embryoid body (EB) and rat engineered heart tissue (EHT).
Ivabradine hydrochloride has been used to investigate the effects of ivabradine on human primary chondrocytes (HPCs) to explore its potential in the treatment of osteoarthritis (OA).

Biochem/physiol Actions

Ivabradine hydrochloride is an open channel blocker of neuronal I(h) and related cardiac I(f) channels, which are comprised of subunits of the hyperpolarisation-activated cyclic nucleotide-gated (HCN) channels, in particular HCN4. Ivabradine has antianginal activity. It lowers heart rate and reduces myocardial oxygen demand.
Ivabradine hydrochloride is an open channel blocker of neuronal I(h) and related cardiac I(f) channels; HCN4 channel blocker; antianginal.
Ivabradine is used to treat chronic heart failure.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Cyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Zhang et al.
The Journal of pharmacology and experimental therapeutics, 368(1), 50-58 (2018-11-10)
An injury of the somatosensory system causes neuropathic pain, which is usually refractory to conventional analgesics, thus warranting the development of novel drugs against this kind of pain. The mechanism of neuropathic pain in rats that had undergone left L5
Elka Popova et al.
Acta neurobiologiae experimentalis, 80(2), 192-204 (2020-07-01)
Hyperpolarization‑activated and cyclic nucleotide‑gated (HCN) channels are well expressed in the vertebrate retina. Their role in formation of electroretinographic (ERG) responses to stimulus onset (b‑wave) and stimulus offset (d‑wave) are largely unknown. In this study we investigated the effects of
Ivabradine abrogates TNF-alpha-induced degradation of articular cartilage matrix
Xiang X, et al.
International Immunopharmacology, 66, 347-353 (2019)
Ivabradine as adjuvant treatment for chronic heart failure
Stoppe C, et al.
Cochrane database of systematic reviews (Online), 2018(4) (2018)
Xianxiang Xiang et al.
International immunopharmacology, 66, 347-353 (2018-12-07)
Ivabradine is most commonly used for the treatment of worsening cardiac failure in patients who cannot tolerate the maximum dose of β-blockers or in whom treatment with β-blockers is contraindicated. While ivabradine is regarded as a highly selective "funny current"

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service