Skip to Content

C5366

Chlorisondamine diiodide

≥98% (HPLC), nicotinic acetylcholine receptor antagonist, solid

Synonym(s):

4,5,6,7-Tetrachloro-2,3-dihydro-2-methyl-2-[2-(trimethylammonio)ethyl]-2H-isoindolium diiodide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H20Cl4I2N2
CAS Number:
Molecular Weight:
611.94
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chlorisondamine diiodide, ≥98% (HPLC), white, solid

Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

white

solubility

H2O: ≥2 mg/mL, DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

[I-].[I-].C[N+](C)(C)CC[N+]1(C)Cc2c(Cl)c(Cl)c(Cl)c(Cl)c2C1

InChI

1S/C14H20Cl4N2.2HI/c1-19(2,3)5-6-20(4)7-9-10(8-20)12(16)14(18)13(17)11(9)15;;/h5-8H2,1-4H3;2*1H/q+2;;/p-2

InChI key

FPNVAOZHQUJJJQ-UHFFFAOYSA-L

Application

Chlorisondamine diiodide has been used:
  • as a nicotinic receptor antagonist to test its effect on trinitrobenzene sulfonic acid (TNBS)-induced colitis
  • as an irreversible nicotinic acetylcholine(nAChR) blocker to pre-treat brain samples to test its effect on cytochrome P450 2B (CYP2B) induction
  • as a ganglionic blocker to test its effect on regulating corticosterone levels in rat with chronic stress

Biochem/physiol Actions

Chlorisondamine diiodide mediates ganglionic and central blockade.
Irreversible, long-lasting nicotinic acetylcholine receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library