Skip to Content
Merck
All Photos(1)

Key Documents

P7791

Sigma-Aldrich

Prazosin hydrochloride

≥99.0% (HPLC), powder, α1-adrenoceptor antagonist

Synonym(s):

1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine hydrochloride, Furazosin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N5O4 · HCl
CAS Number:
Molecular Weight:
419.86
Beilstein:
4303561
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Prazosin hydrochloride, ≥99.0% (HPLC)

Assay

≥99.0% (HPLC)

form

powder

solubility

H2O: 0.5 mg/mL
methanol: 6 mg/mL
dilute aqueous acid: insoluble

SMILES string

Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)c4ccco4

InChI

1S/C19H21N5O4.ClH/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14;/h3-4,9-11H,5-8H2,1-2H3,(H2,20,21,22);1H

InChI key

WFXFYZULCQKPIP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Prazosin is a member of the class of α-adrenergic blockers. It is considered as a peripheral vasodilator antihypertensive agent. Prazosin is a derivative of quinazoline.

Application

Prazosin hydrochloride has been used:
  • to block the α1adrenergic receptors that mediate sympathetic vasoconstriction in mice
  • as an α1-adrenoceptor blocker,administered intragastrically in rats
  • as a vasodilator,administered together with inuslin into the left ventricle of mice for the assessment of its effects on renal functions

Biochem/physiol Actions

Peripheral α1-adrenoceptor antagonist; vasodilator. Prazosin can be used to treat chronic posttraumatic stress disorder (PTSD). It plays a role in reducing blood pressure by relaxing blood vessels, hence used in the treatment of high blood pressure.

Features and Benefits

This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the α1-Adrenoceptors, α2-Adrenoceptors and Melatonin Receptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Hygroscopic; light sensitive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nia C S Lewis et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 304(2), R147-R154 (2012-11-24)
We examined the hypothesis that α(1)-adrenergic blockade would lead to an inability to correct initial orthostatic hypotension (IOH) and cerebral hypoperfusion, leading to symptoms of presyncope. Twelve normotensive humans (aged 25 ± 1 yr; means ± SE) attempted to complete
Aslak Wøien Augedal et al.
Sleep medicine reviews, 17(2), 143-152 (2012-10-11)
A meta-analysis of treatments for nightmares is reported. The studies were identified by database searches and by an inspection of relevant reference lists. The inclusion criteria were: nightmares as a target problem, studies published in English, use of a randomized
Quantitative analysis of prazosin hydrochloride in pharmaceutical formulation by prazosin potentiometric sensor based on computational investigation
Faridbod F, et al.
International Journal of Electrochemical Science, 5(4), 653-667 (2010)
Enhanced salt sensitivity following shRNA silencing of neuronal TRPV1 in rat spinal cord
Yu S Q and Wang D H
Acta Pharmacologica Sinica, 32(6), 845-845 (2011)
Haptoglobin reduces renal oxidative DNA and tissue damage during phenylhydrazine-induced hemolysis
Lim Y K, et al.
Kidney International, 58(3), 1033-1044 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service