Skip to Content
Merck
All Photos(2)

Key Documents

U101

Sigma-Aldrich

5-Methylurapidil

solid

Synonym(s):

5-Methyl-6[[3-[4-(2-methoxyphenyl)-1-piperazinyl]­propyl]­amino]-1,3-dimethyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31N5O3
CAS Number:
Molecular Weight:
401.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

H2O: 0.4 mg/mL
0.1 M HCl: 3.8 mg/mL

SMILES string

Cl[H].COc1ccccc1N2CCN(CCCNC3=C(C)C(=O)N(C)C(=O)N3C)CC2

InChI

1S/C21H31N5O3.ClH/c1-16-19(23(2)21(28)24(3)20(16)27)22-10-7-11-25-12-14-26(15-13-25)17-8-5-6-9-18(17)29-4;/h5-6,8-9,22H,7,10-15H2,1-4H3;1H

InChI key

WAZDYFHTLYHMKO-UHFFFAOYSA-N

Gene Information

Application

5-Methylurapidil has been used for competitive binding in radioligand binding assays.

Biochem/physiol Actions

Selective α1A-adrenoceptor antagonist; antihypertensive.

Features and Benefits

This compound is featured on the α1-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

5-Methylurapidil is soluble in water at 0.4 mg/ml and is also soluble in 0.1 M HCl at 3.8 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Endoh et al.
Naunyn-Schmiedeberg's archives of pharmacology, 345(5), 578-585 (1992-05-01)
In order to elucidate the contribution of alpha 1A subtype to the positive inotropic effect mediated by myocardial alpha 1 adrenoceptors, the influence of the alpha 1A selective antagonists WB 4101 and 5-methylurapidil on the alpha 1-mediated positive inotropic effect
Caroline Putzke et al.
Cardiovascular research, 75(1), 59-68 (2007-03-29)
The outward current flowing through the two-pore domain acid-sensitive potassium channel TASK-1 (I(TASK)) and its inhibition via alpha1-adrenergic receptors was studied in rat ventricular cardiomyocytes. Quantitative RT-PCR experiments were carried out with mRNA from rat heart. Patch-clamp recordings were performed
Rocío Alcántara-Hernández et al.
Biochimica et biophysica acta, 1864(12), 2378-2388 (2017-09-11)
In LNCaP cells that stably express α
Abdul H Khan et al.
European journal of pharmacology, 569(1-2), 110-118 (2007-06-15)
This study investigated whether the alpha(1)-adrenoceptor subtype(s) mediating the vasoconstrictor actions of the renal sympathetic nerves were altered in rats with cisplatin-induced renal failure. Male Wistar Kyoto rats were used and half received cisplatin (5 mg/kg i.p.) to induce renal
Wesley B Asher et al.
Journal of chemical information and modeling, 47(5), 1906-1912 (2007-08-25)
In this study, we have developed a two model system to mimic the active and inactive states of a G-protein coupled receptor specifically the alpha1A adrenergic receptor. We have docked two agonists, epinephrine (phenylamine type) and oxymetazoline (imidazoline type), as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service