Skip to Content
Merck
All Photos(2)

Key Documents

S7395

Sigma-Aldrich

Spiperone

solid

Synonym(s):

8-[3-(p-Fluorobenzoyl)propyl]-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one, R 5147, Spiroperidol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26FN3O2
CAS Number:
Molecular Weight:
395.47
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

light yellow

solubility

H2O: slightly soluble 0.2 mg/mL
0.1 M HCl: slightly soluble 0.3 mg/mL
ethanol: 1.5 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2 mg/mL

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCC3(CC2)N(CNC3=O)c4ccccc4

InChI

1S/C23H26FN3O2/c24-19-10-8-18(9-11-19)21(28)7-4-14-26-15-12-23(13-16-26)22(29)25-17-27(23)20-5-2-1-3-6-20/h1-3,5-6,8-11H,4,7,12-17H2,(H,25,29)

InChI key

DKGZKTPJOSAWFA-UHFFFAOYSA-N

Related Categories

General description

Spiperone is a butyrophenone antipsychotic agent. It induces calcium-dependent chloride secretion in the airway and functions as a potential therapeutic target for cystic fibrosis.

Application

Spiperone has been used to block the actions of 5-hydroxytryptamine (5-HT) receptor. It also has been used to study its anti tumor effects in glioblastoma (GBM) cell lines.
Spiperone was used to study the role of dopamine receptors in facilitating the male sexual behavior in quails.6

Biochem/physiol Actions

Selective D2 dopamine receptor antagonist; α1B-adrenoceptor antagonist; mixed 5-HT2A/5-HT1 serotonin receptor antagonist; antipsychotic.

Reconstitution

Solutions may be stored for 1-2 days at 4 °C.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I S Geerts et al.
British journal of pharmacology, 127(6), 1327-1336 (1999-08-24)
In humans intimal thickening is aprerequisite of atherosclerosis. Application of a silicone collar around the rabbit carotid artery induces an intimal thickening but in addition it increases the sensitivity to the vasoconstrictor action of serotonin (5-hydroxytryptamine, 5-HT). The 5-HT receptors
Spiperone, identified through compound screening, activates calcium-dependent chloride secretion in the airway
Liang L, et al.
American Journal of Physiology. Cell Physiology, 296(1), C131-C141 (2009)
Marion Schuller et al.
ChemMedChem, 12(19), 1585-1594 (2017-08-05)
MS Binding Assays are a label-free alternative to radioligand binding assays. They provide basically the same capabilities as the latter, but use a non-labeled reporter ligand instead of a radioligand. In contrast to radioligand binding assays, MS Binding Assays offer-owing
Carmen Klein Herenbrink et al.
Nature communications, 7, 10842-10842 (2016-02-26)
Biased agonism describes the ability of ligands to stabilize different conformations of a GPCR linked to distinct functional outcomes and offers the prospect of designing pathway-specific drugs that avoid on-target side effects. This mechanism is usually inferred from pharmacological data
Spiperone enhances intracellular calcium level and inhibits the Wnt signaling pathway
Lu D and Carson DA
BioMed Central Pharmacology, 9(1), 13-13 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service