Skip to Content
Merck
All Photos(5)

Key Documents

T9534

Sigma-Aldrich

Triethanolamine hydrochloride

BioXtra, ≥99.5% (titration)

Synonym(s):

2-[Bis(2-hydroxyethyl)amino]ethanol hydrochloride, TEA-Hydrochloride, trolamine hydrochloride, 2,2′,2′′-Nitrilotriethanol hydrochloride, Tris(2-hydroxyethyl)amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N · HCl
CAS Number:
Molecular Weight:
185.65
Beilstein:
3909940
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99.5% (titration)

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

useful pH range

7.3-8.3

pKa (25 °C)

7.8

mp

177-179 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

diagnostic assay manufacturing

SMILES string

Cl.OCCN(CCO)CCO

InChI

1S/C6H15NO3.ClH/c8-4-1-7(2-5-9)3-6-10;/h8-10H,1-6H2;1H

InChI key

HHLJUSLZGFYWKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethanolamine hydrochloride (TEA-HCl) is a salt form of triethanolamine and is commonly used as a buffer substance in biochemistry. It aids in maintaining the stable pH level in a solution during biochemical reactions. TEA-HCL serves as a buffering agent during protein extraction and protein purification. It is commonly used to adjust the pH of buffer solutions in enzyme assays. In addition to its buffering property, it also exhibits surfactant and emulsifying properties and can be used in some cosmetic and personal care products to adjust the pH and improve solubility.

Application

Triethanolamine hydrochloride buffer has been used in in-situ mRNA hybridization for acetylation of frozen tissues mounted on slides.

Features and Benefits

  • The BioXtra product line ensures high-quality products for consistent results.
  • A high assay of ≥99.5% guarantees the purity of the product.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasticity of endothelial cells: rapid dedifferentiation of freshly isolated high endothelial venule endothelial cells outside the lymphoid tissue microenvironment
Lacorre D A, et al.
Blood, 103(11), 4164- 472 (2004)
Enzymatic catalysis of proton transfer at carbon: activation of triosephosphate isomerase by phosphite dianion
Amyes T L and Richard J P
Biochemistry, 46(19) , 5841-554 (2007)
Assaying plant formate-tetrahydrofolate ligase with monoglutamylated and polyglutamylated substrates using a fluorescence-HPLC based method
Saeheng S and Roje S
Methods in Enzymology, 680 (2022)
Formulation effects of topical emulsions on transdermal and dermal delivery
Otto A, et al.
International Journal of Cosmetic Science, 31(1) , 1-19 (2009)
The rough endoplasmic reticulum-resident FK506-binding protein FKBP65 is a molecular chaperone that interacts with collagens
Ishikawa Y, et al.
The Journal of Biological Chemistry, 283(46) , 31584-31590 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service