Skip to Content
Merck
All Photos(1)

Key Documents

90350

Sigma-Aldrich

Triethylamine hydrochloride

≥99.0% (AT)

Synonym(s):

N,N-Diethylethanamine hydrochloride, Triethylamine monohydrochloride, Triethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N · HCl
CAS Number:
Molecular Weight:
137.65
Beilstein:
3906409
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99.0% (AT)

form

crystals

mp

261 °C (dec.) (lit.)

solubility

water: soluble 1 g/10 mL

functional group

amine

SMILES string

Cl.CCN(CC)CC

InChI

1S/C6H15N.ClH/c1-4-7(5-2)6-3;/h4-6H2,1-3H3;1H

InChI key

ILWRPSCZWQJDMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Triethylamine hydrochloride can be used as a:
  • Catalyst to synthesize diphenylmethane via Friedel-Crafts reaction of benzene with benzyl chloride.[1]
  • Reactant in the ionic liquid-catalyzed synthesis of quaternary ammonium salts with dialkyl carbonates.[2]
It can also be used as a proton donor:
  • For the titanocene (III)-catalyzed intramolecular ring-opening arylation of epoxides.[3]
  • In intramolecular Kinugasa reaction to synthesize β-lactams.[4][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L De La Rocha et al.
Analytical chemistry, 68(21), 3746-3750 (1996-11-01)
A procedure for the purification, recovery, and determination of isotopic abundances of silicon from biogenic and lithogenic particulate matter and dissolved silicic acid is reported. Purification involves the reaction of acid molybdate with dissolved silicon in natural waters or that
Synthesis of beta-lactams via diastereoselective, intramolecular Kinugasa reactions
Popik O, et al.
Organic & Biomolecular Chemistry, 18, 2852-2860 (2020)
Isabel Rodríguez et al.
International journal of pharmaceutics, 529(1-2), 168-177 (2017-06-28)
This study investigates how both bioadhesive polymers (chitosan, hyaluronic acid and alginate) and permeability enhancers (ethylene glycol- bis(2-aminoethylether)- N, N, N', N'- tetraacetic acid (EGTA) and hydroxypropyl-ß-cyclodextrin) influence the permeability of the anti-glaucoma drug timolol maleate through ex vivo bovine
Hao Chen et al.
Science advances, 3(12), eaao7233-eaao7233 (2017-12-20)
Rechargeable aluminum-ion batteries are promising in high-power density but still face critical challenges of limited lifetime, rate capability, and cathodic capacity. We design a "trihigh tricontinuous" (3H3C) graphene film cathode with features of high quality, orientation, and channeling for local
THE SPECIFIC PRECIPITATION OF ORTHOPHOSPHATE AND SOME BIOCHEMICAL APPLICATIONS.
Y SUGINO et al.
The Journal of biological chemistry, 239, 2360-2364 (1964-07-01)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service