Skip to Content
Merck
All Photos(2)

Key Documents

258148

Sigma-Aldrich

Hydrochloric acid

Hydrochloric acid
1 of 2 reviewers received a sample product or took part in a promotion

37%, ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Hydrochloric acid, ACS reagent, 37%

grade

ACS reagent

Agency

suitable for GB 5009.253-2016
suitable for SM 5210

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

Assay

36.5-38.0% (ACS specification)
37%

form

liquid

impurities

≤1 ppm free chlorine

ign. residue

≤5 ppm

color

APHA: ≤10, free from suspended matter or sediment

pH

<1

bp

>100 °C (lit.)

density

1.18 g/L at 20 °C

anion traces

bromide (Br-): ≤0.005%
sulfate (SO42-): ≤1 ppm
sulfite (SO32-): ≤1 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
NH4+: ≤3 ppm
heavy metals: ≤1 ppm (by ICP)

storage temp.

room temp

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

Single 2.5L-GL units require a can opener to open the metal shipping container.

Other Notes

ACS specifications same as product 320331

Legal Information

OXONE is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Detection of nascent RNA, single-copy DNA and protein localization by immunoFISH in mouse germ cells and preimplantation embryos.
Namekawa SH and Lee JT.
Nature Protocols, 6(3), 270-284 (2011)
Falk Bernsmann et al.
Langmuir : the ACS journal of surfaces and colloids, 27(6), 2819-2825 (2011-02-22)
The deposition of "polydopamine" films, from an aqueous solution containing dopamine or other catecholamines, constitutes a new and versatile way to functionalize solid-liquid interfaces. Indeed such films can be deposited on almost all kinds of materials. Their deposition kinetics does
Satoshi H Namekawa et al.
Nature protocols, 6(3), 270-284 (2011-03-05)
Dynamic reprogramming of the genome takes place during the gamete-to-embryo transition. This transition defines a period of continuous and global change but has been difficult to study because of extremely limited material and varying degrees of chromatin compaction. Improved methods
Eagleson M.
Concise Encyclopedia Chemistry, 52-52 (1994)
Zhibo Liu et al.
Nature protocols, 10(9), 1423-1432 (2015-08-28)
Herein we present a general protocol for the functionalization of biomolecules with an organotrifluoroborate moiety so that they can be radiolabeled with aqueous (18)F fluoride ((18)F(-)) and used for positron emission tomography (PET) imaging. Among the β(+)-emitting radionuclides, fluorine-18 ((18)F)

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is this HCl dilluted in ETOH?

    1 answer
    1. According to Section 15 of the product Safety Data Sheet, this product is diluted in water.

      Helpful?

  4. Hi! What concentration does this solution have in mol/dm^3? Kind regards

    1 answer
    1. Molarity (moles/L) = Density (g/L) ÷ Molecular Weight (g/mol) * purity
      = 1200 g/L ÷ 36.46 g/mol * 0.373
      = 12.276 mol/L

      Helpful?

  5. What is the difference between 258148-6x500ml and 25148-6x500ml-GL? I want the ones that come in non-glass bottles.

    1 answer
    1. Product 258148-6X500ML is 6 bottles with 500 mL of HCl in a plastic bottle. The SKU ending with the -GL signifies the glass bottle. The difference is in the packaging of the reagent.

      Helpful?

Reviews

1 of 2 reviewers received a sample product or took part in a promotion

Active Filters

    • Review 1
    • Votes 31
    5 out of 5 stars.

    HCL

    Very nice, very acidic. Would buy again. High satisfaction.

    Helpful?

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service