Skip to Content
Merck
All Photos(2)

Key Documents

T7313

Sigma-Aldrich

1-[2-(Trifluoromethyl)phenyl]imidazole

Synonym(s):

TRIM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F3N2
CAS Number:
Molecular Weight:
212.17
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (GC)

form

solid

solubility

DMSO: soluble 30 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ccccc1-n2ccnc2

InChI

1S/C10H7F3N2/c11-10(12,13)8-3-1-2-4-9(8)15-6-5-14-7-15/h1-7H

InChI key

WZBWBNCQUTXYEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent nitric oxide synthase inhibitor. Because of the putative involvement of nitric oxide in depression and stress syndromes, TRIM was studied for its antidepressant effects, and found to reduce manifestations of stress in an animal model as well as fluoxetine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amélie Wallenhammar et al.
PloS one, 12(7), e0181551-e0181551 (2017-07-29)
Tripartite motif-containing (TRIM) proteins are defined by the sequential arrangement of RING, B-box and coiled-coil domains (RBCC), where the B-box domain is a unique feature of the TRIM protein family. TRIM21 is an E3 ubiquitin-protein ligase implicated in innate immune
Yves Basset et al.
Ecology and evolution, 7(23), 9991-10004 (2017-12-15)
We have little knowledge of the response of invertebrate assemblages to climate change in tropical ecosystems, and few studies have compiled long-term data on invertebrates from tropical rainforests. We provide an updated list of the 72 species of Saturniidae moths
Juan Traba et al.
Scientific reports, 9(1), 9473-9473 (2019-07-03)
Farmland bird populations have strongly declined across Europe over the last decades due to agriculture intensification, despite successive reforms of EU's Common Agricultural Policy (CAP). In parallel, CAP has led to a reduction of fallow land, a critical habitat for
R L Handy et al.
British journal of pharmacology, 119(2), 423-431 (1996-09-01)
1. The ability of a range of substituted imidazole compounds to inhibit mouse cerebellar neuronal nitric oxide synthase (nNOS), bovine aortic endothelial NOS (eNOS) and inducible NOS (iNOS) from lungs of endotoxin-pretreated rats was investigated. In each case the substrate
Eva Šatović et al.
Scientific reports, 9(1), 19962-19962 (2019-12-29)
Terminal repeat retrotransposons in miniature (TRIMs) are small non-autonomous LTR retrotransposons consisting of two terminal direct repeats surrounding a short internal domain. The detection and characterization of these elements has been mainly limited to plants. Here we present the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service