Skip to Content
Merck
All Photos(1)

Key Documents

T3757

Sigma-Aldrich

TTNPB

Synonym(s):

4-[(E)-2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid, Arotinoid acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28O2
CAS Number:
Molecular Weight:
348.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

solubility

ethanol: soluble 10 mM
DMSO: soluble 25 mM
chloroform/methanol: soluble 9.80-10.20 mg/mL, clear, colorless to light yellow

Quality Level

storage temp.

−20°C

SMILES string

C\C(=C/c1ccc(cc1)C(O)=O)c2ccc3c(c2)C(C)(C)CCC3(C)C

InChI

1S/C24H28O2/c1-16(14-17-6-8-18(9-7-17)22(25)26)19-10-11-20-21(15-19)24(4,5)13-12-23(20,2)3/h6-11,14-15H,12-13H2,1-5H3,(H,25,26)/b16-14+

InChI key

FOIVPCKZDPCJJY-JQIJEIRASA-N

Application

TTNPB has been used for transcriptional assays in 293T cells4. It has also been used as a RAR-agonist in cultured human cord blood CD34+CD38-lin- cells5.

Biochem/physiol Actions

Selective and highly potent retinoic acid analog with affinity for retinoic acid receptors (RAR) α, β, and γ, which are nuclear transcription factors. Produces ligand-activated transcription of genes that possess retinoic acid responsive elements.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

TTNPB dissolves in CHCL3/MeOH (1/1) at 9.80 - 10.20 mg/ml to yield a clear, colorless to light yellow solution. It is also soluble at 10 mM in ethanol and at 25 mM in DMSO.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias W Büttner et al.
Chembiochem : a European journal of chemical biology, 8(14), 1688-1699 (2007-09-05)
Twofold sila-substitution (C/Si exchange) in the saturated ring of the tetrahydronaphthalene skeleton of the retinoid agonists TTNPB (1 a) and 3-methyl-TTNPB (2 a) leads to disila-TTNPB (1 b) and disila-3-methyl-TTNPB (2 b), respectively. The silicon compounds 1 b and 2
Bryan R Haugen et al.
The Journal of clinical endocrinology and metabolism, 89(1), 272-280 (2004-01-13)
Therapy for patients with advanced thyroid carcinoma is limited. Clinical and in vitro studies suggest that some patients with advanced thyroid cancer may respond to therapy with retinoic acid. mRNA expression of the six retinoic acid (RAR) and retinoid X
Olivier M Niemoeller et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 21(1-3), 193-202 (2008-01-23)
Vitamin A and retinoic acid have previously been shown to confer some protection against a severe course of malaria by fostering the phagocytosis of parasitized erythrocytes. Phagocytosis of erythrocytes is stimulated by phosphatidylserine exposure at the cell surface. The present
M S Sheikh et al.
The Journal of biological chemistry, 269(34), 21440-21447 (1994-08-26)
Retinoids mediate their actions via RARs (retinoic acid receptors) and RXRs (retinoid X receptors). Each class of these nuclear retinoid receptors is further subdivided into three species, namely alpha, beta, and gamma. Recent studies demonstrate that estrogen receptor (ER)-positive human
Masakazu Sato et al.
Oncotarget, 8(25), 40935-40945 (2017-04-14)
Cervical reserve cells are epithelial progenitor cells that are pathologically evident as the origin of cervical cancer. Thus, investigating the characteristics of cervical reserve cells could yield insight into the features of cervical cancer stem cells (CSCs). In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service