Skip to Content
Merck
All Photos(1)

Key Documents

SML0282

Sigma-Aldrich

Bexarotene

≥98% (HPLC)

Synonym(s):

4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid, LGD-1069, SR-11247

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28O2
CAS Number:
Molecular Weight:
348.48
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥15 mg/mL (warmed)

storage temp.

−20°C

SMILES string

C=C(C1=CC=C(C(O)=O)C=C1)C2=C(C)C=C3C(C(C)(C)CCC3(C)C)=C2

InChI

1S/C24H28O2/c1-15-13-20-21(24(5,6)12-11-23(20,3)4)14-19(15)16(2)17-7-9-18(10-8-17)22(25)26/h7-10,13-14H,2,11-12H2,1,3-6H3,(H,25,26)

InChI key

NAVMQTYZDKMPEU-UHFFFAOYSA-N

Gene Information

Application

Bexarotene has been used as retinoid x receptor (RXR) agonist:
  • to study its effect on human oral squamous cell carcinoma (OSCC) cell lines
  • to test for cooperative cell killing of cutaneous T cell lymphoma (CTCL) with 966
  • as a standard in HPLC for determining its potential impurities in drug substances and drug products

Bexarotene has been used:
  • to investigate its effects on the changes in renal, cardiac, hepatic, and pulmonary expression/activity of inducible nitric oxide synthase (iNOS) and CYP4F6
  • to analyze the involvement of retinoid x receptor (RXR) homo or permissive heterodimers
  • to study its potential impurities in drug substances and drug products

Biochem/physiol Actions

Bexarotene is a highly selective retinoid X receptor (RXR) agonist and an antineoplastic agent.
Bexarotene is a highly selective retinoid X receptor (RXR) agonist. It is an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer′s model lowered the most toxic form of β-amyloid peptide and increased cognitive ability. The activity in the mouse Alzheimer′s models are believed to be by activating PPARγ:RXR and LXR:RXR dimers which induces the expression of apoE and facilitates Aβ clearance and promotes microglial phagocytosis.
Bexarotene is used to treat breast cancer.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shawn C Owen et al.
ACS chemical biology, 7(8), 1429-1435 (2012-05-26)
Many small molecules, including bioactive molecules and approved drugs, spontaneously form colloidal aggregates in aqueous solution at micromolar concentrations. Though it is widely accepted that aggregation leads to artifacts in screens for ligands of soluble proteins, the effects of colloid
Protective effects of retinoid x receptors on retina pigment epithelium cells
Ayala-Pena VB, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1863(6), 1134-1145 (2016)
Retinoids induce antagonism between FOXO3A and FOXM1 transcription factors in human oral squamous cell carcinoma (OSCC) cells
Osei-Sarfo K, et al.
PLoS ONE, 14(4), e0215234-e0215234 (2019)
Bexarotene, a Selective RXRalpha Agonist, Reverses Hypotension Associated with Inflammation and Tissue Injury in a Rat Model of Septic Shock
Tunctan B, et al.
Inflammation, 41(1), 337-355 (2018)
The rexinoid bexarotene represses cyclin D1 transcription by inducing the DEC2 transcriptional repressor
Li Y, et al.
Breast Cancer Research and Treatment, 128(3), 667-677 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service