Skip to Content
Merck
All Photos(1)

Documents

P1269

Sigma-Aldrich

Phorbol 12,13-dibutyrate

≥98% (TLC), powder, PKC activator

Synonym(s):

PDBu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40O8
CAS Number:
Molecular Weight:
504.61
Beilstein:
6551234
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

product name

Phorbol 12,13-dibutyrate, ≥98% (TLC), powder

Quality Level

Assay

≥98% (TLC)

form

powder

color

clear light yellow, film

solubility

H2O: 30 μM
DMSO: soluble
acetone: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

CCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)[C@@H](C=C(CO)C[C@@]3(O)[C@H]2C=C(C)C3=O)[C@@H]4C(C)(C)[C@]14OC(=O)CCC

InChI

1S/C28H40O8/c1-7-9-20(30)35-24-16(4)27(34)18(22-25(5,6)28(22,24)36-21(31)10-8-2)12-17(14-29)13-26(33)19(27)11-15(3)23(26)32/h11-12,16,18-19,22,24,29,33-34H,7-10,13-14H2,1-6H3/t16-,18+,19-,22-,24-,26-,27-,28-/m1/s1

InChI key

BQJRUJTZSGYBEZ-YVQNUNKESA-N

Gene Information

mouse ... Prkcd(18753)

Looking for similar products? Visit Product Comparison Guide

Application

Phorbol 12,13-dibutyrate has been used as a protein kinase C (PKC) activator to study its effects on human organic anion transporter 4 (hOAT4) activity. It has also been used as a PKC activator to analyze its effects on galectin-3 expression and matrix production in HL-1 cells.

Biochem/physiol Actions

Phorbol 12,13-dibutyrate (PDBu) is a potent activator of protein kinase C (PKC). It is more hydrophilic than phorbol 12-myristate 13-acetate (PMA), which facilitates washing PDBu out of cells in tissue culture. PDBu activates endothelial nitric oxide synthase expression in primary human umbilical vein endothelial cells, which in turn correlates with PKC α and ε expression.

Features and Benefits

Less hydrophobic then PMA, making it easier to wash out of cells in tissue culture.

Caution

Photosensitive

Reconstitution

Stock solution cannot be made in aqueous media; dissolve in a water-miscible organic solvent before dilution to working concentrations in aqueous media.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quynh Nhu Dinh et al.
Aging, 9(6), 1595-1606 (2017-07-01)
Aging is commonly associated with chronic low-grade inflammation and hypertension but it is unknown whether a cause-effect relationship exists between them. We compared the sensitivity of young adult (8-12 w) and aged (23-31 mo) male C57Bl6J mice to develop hypertension
Vesna Lazarevic et al.
International journal of molecular sciences, 20(17) (2019-09-05)
Major depressive disorder is one of the most common neuropsychiatric disorders worldwide. The treatment of choice that shows good efficacy in mood stabilization is based on selective serotonin reuptake inhibitors (SSRIs). Their primary mechanism of action is considered to be
T Hori et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(17), 7262-7267 (1999-08-25)
Phorbol ester facilitates transmitter release at a variety of synapses, and the phorbol ester-induced synaptic potentiation (PESP) is a model for presynaptic facilitation. To address the mechanism underlying PESP, we have made paired whole-cell recordings from the giant presynaptic terminal
Mirjana Efremova et al.
Nature communications, 9(1), 32-32 (2018-01-04)
The cancer immunoediting hypothesis postulates a dual role of the immune system: protecting the host by eliminating tumor cells, and shaping the tumor by editing its genome. Here, we elucidate the impact of evolutionary and immune-related forces on editing the
Kazuhiro Irie et al.
Pharmacology & therapeutics, 93(2-3), 271-281 (2002-08-23)
Conventional and novel protein kinase C (PKC) isozymes contain two cysteine-rich C1 domains (C1A and C1B), both of which are candidate phorbol-12, 13-dibutyrate (PDBu)-binding sites. We synthesized C1 peptides of 50-70 residues corresponding to all PKC isozyme C1 domains using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service