Skip to Content
Merck
All Photos(2)

Key Documents

SML0305

Sigma-Aldrich

3-Deazaneplanocin A hydrochloride

≥97% (HPLC)

Synonym(s):

(-)-1-[(1R,4R,5S)-3-(Hydroxymethyl)-4,5-dihydroxy-2-cyclopenten-1-yl]4-aminoimidazo[4,5-c]pyridine hydrochloride, DZNep hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O3 · HCl
CAS Number:
Molecular Weight:
298.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D -88 to -108°, c = 0.1 in water

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

Cl.Nc1nccc2n(cnc12)[C@@H]3C=C(CO)[C@@H](O)[C@H]3O

InChI

1S/C12H14N4O3.ClH/c13-12-9-7(1-2-14-12)16(5-15-9)8-3-6(4-17)10(18)11(8)19;/h1-3,5,8,10-11,17-19H,4H2,(H2,13,14);1H/t8-,10-,11+;/m1./s1

InChI key

UNSKMHKAFPRFTI-FDKLLANESA-N

General description

3-Deazaneplanocin A hydrochloride is a carbocyclic analog of adenosine. It has antimetastatic property. It is used to treat Ebola virus disease, gastric cancer, myeloma and lymphoma.

Application

3-Deazaneplanocin A hydrochloride has been used to examine the potential involvement of autophagy and apoptosis in colorectal cancer (CRC) cells.
3-Deazaneplanocin A hydrochloride has been used to test its effect on ovarian tumor progression. It has also been used as a S-adenosyl homocysteine hydrolase (SAH) inhibitor in c33A2 cells.

Biochem/physiol Actions

3-Deazaneplanocin A (DZNep) is an S-Adenosylhomocysteine Hydrolase inhibitor and histone methyltransferase EZH2 inhibitor. It switches on repressed tumor suppressor genes and induces apoptosis by inhibiting the expression of EZH2.
3-Deazaneplanocin A is an S-Adenosylhomocysteine Hydrolase inhibitor and histone methyltransferase EZH2 inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-Deazaneplanocin A (DZNep), an inhibitor of S-adenosylmethionine-dependent methyltransferase, promotes erythroid differentiation
Fujiwara T, et al.
The Journal of Biological Chemistry, 289(12), 8121-8134 (2014)
Song-Hee Kim et al.
Biomolecules, 11(4) (2021-05-01)
EPHA3, a member of the EPH family, is overexpressed in various cancers. We demonstrated previously that EPHA3 is associated with radiation resistance in head and neck cancer via the PTEN/Akt/EMT pathway; the inhibition of EPHA3 significantly enhances the efficacy of
Epigenetic silencing of T H 1-type chemokines shapes tumour immunity and immunotherapy
Peng D, et al.
Nature, 527(7577), 249-249 (2015)
Chengfa Zhao et al.
Reproduction (Cambridge, England), 157(4), 359-369 (2019-02-08)
Somatic cell nuclear transfer in mammalian cloning suffers from a faulty epigenetic reprogramming, which is believed to cause developmental failures in cloned embryos. Regulating the epigenetic-modifying enzymes can rescue the chromatin of cloned embryos from aberrant epigenetic status, thereby potentially
Downregulation of enhancer of Zeste Homolog 2 (EZH2) is essential for the induction of autophagy and apoptosis in colorectal cancer cells
Yao Y, et al.
Genes, 7(10), 83-83 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service