Skip to Content
Merck
All Photos(1)

Key Documents

T0251

Sigma-Aldrich

Thymidine 5′-triphosphate sodium salt

≥96%

Synonym(s):

TTP, dTTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O14P3Na
CAS Number:
Molecular Weight:
504.15
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥96%

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NaH].[P](=O)(O[P](=O)(OCC1OC(CC1O)N2C=C(C(=O)NC2=O)C)O)(O[P](=O)(O)O)O

InChI

1S/C10H17N2O14P3.Na.H/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(24-8)4-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,21,22)(H,11,14,15)(H2,16,17,18);;

InChI key

KAELIXTVSANDSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thymidine 5′-triphosphate (dTTP) is one of the four natural deoxynucleotides, along with deoxyadenosine 5′-triphosphate (dATP), deoxyguanosine 5′-triphosphate (dGTP) and deoxycytosine 5′-triphosphate (dCTP) used for the biosynthesis of deoxyribonucleic acids by DNA polymerases and reverse transcriptases. Deoxy-TTP may also be used as a substrate to study the specificity and kinetics of thymidine-triphosphatase(s) (EC 3.6.1.39) and other dTTP hydrolyzing activities.
Thymidine 5′-triphosphate sodium salt has been used:
  • in the quenching ability test for dual-aptamer and multiple-nucleotide detection based on a multiple-aptamer/GO-nS complex
  • to study protein-metabolite interactions in the central metabolism of Escherichia coli
  • in a polymerase chain reaction (PCR) product formation using AM-toxin primers

Biochem/physiol Actions

Thymidine triphosphate is a building block of DNA. It acts as an allosteric regulator of other nucleotides′ metabolism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interconversion of deoxyribonucleotides
Henderson JF and A.R.P.Paterson
Nucleosides, 247-259 (2018)
Hidenori Nakaoka et al.
The Journal of biological chemistry, 287(1), 619-627 (2011-11-17)
The Ctc1-Stn1-Ten1 (CST) complex is an RPA (replication protein A)-like protein complex that binds to single-stranded (ss) DNA. It localizes at telomeres and is involved in telomere end protection in mammals and plants. It is also known to stimulate DNA
Christina Jensen et al.
Scientific reports, 7(1), 2830-2830 (2017-06-08)
In the absence of a validated correlate of protection or robust animal models for human tuberculosis, Mycobacterial growth inhibition assays (MGIAs) aim to assess vaccines ability to inhibit mycobacterial growth in-vitro. We optimised a reproducible murine splenocyte MGIA based on
Xue-Ke Zhao et al.
PloS one, 11(12), e0167451-e0167451 (2016-12-03)
Tumor necrosis factor-alpha (TNF-α) is a potent pro-inflammatory mediator and its expression is up-regulated in chronic obstructive pulmonary disease (COPD). Tristetraprolin (TTP) is implicated in regulation of TNF-α expression; however, whether TTP is involved in cigarette smoke-induced TNF-α expression has
Merel M C Lambregts et al.
PloS one, 14(1), e0208819-e0208819 (2019-01-03)
Blood cultures are considered the gold standard to distinguish bacteremia from non-bacteremic systemic inflammation. In current clinical practice, bacteraemia is considered unlikely if blood cultures have been negative for 48-72 hours. Modern BC systems have reduced this time-to-positivity (TTP), questioning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service