Skip to Content
Merck

Skip To

D4010

2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate

≥96% (HPLC), synthetic (organic), powder

Synonym(s):

2′-deoxy-, 5′-triphosphate sodium salt, Guanosine, dGTP sodium salt hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H16N5O13P3 · xNa+ · yH2O
CAS Number:
Molecular Weight:
507.18 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51
PubChem Substance ID:
Assay:
≥96% (HPLC)
Biological source:
synthetic (organic)
Form:
powder
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate, ≥96% (HPLC)

biological source

synthetic (organic)

Quality Segment

assay

≥96% (HPLC)

form

powder

shipped in

dry ice

storage temp.

−20°C

Application

2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate has been used:
  • as a component in a polymerase chain reaction (PCR) mixture for PCR product formation using AM-toxin primers[1]
  • as an internal standard to improve the analytical performance of capillary electrophoresis (CE) analysis for the evaluation of the cleanup process[2]
  • as a putative substrate in in vitro enzyme activity assay to incubate with nudix hydrolase 15 (NUDT15) protein for the evaluation of putative NUDT15 inhibitors[3]

Biochem/physiol Actions

Deoxyguanosine 5′-triphosphate (dGTP), a purine deoxynucleotide, is a substrate of DNA polymerase(s) and reverse transcriptases. dGTP is useful to study the distribution, specificity, and kinetics of various nucleoside-triphosphatases (NTPase) and ribonucleotide reductases.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D7409D5038D7170
biological source

synthetic (organic)

biological source

-

biological source

-

biological source

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

assay

≥96% (HPLC)

assay

≥90% (HPLC)

assay

≥99%

assay

≥99%

form

powder

form

powder

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

dry ice

shipped in

-

shipped in

dry ice

shipped in

dry ice


Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters