Skip to Content
Merck

Skip To

D4635

2′-Deoxycytidine 5′-triphosphate disodium salt

≥95%

Synonym(s):

dCTP-Na2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H14N3Na2O13P3
CAS Number:
Molecular Weight:
511.12
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
MDL number:
Assay:
≥95%
Form:
powder
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95%

form

powder

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O2

InChI

1S/C9H16N3O13P3.2Na/c10-7-1-2-12(9(14)11-7)8-3-5(13)6(23-8)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;/h1-2,5-6,8,13H,3-4H2,(H,18,19)(H,20,21)(H2,10,11,14)(H2,15,16,17);;/q;2*+1/p-2/t5-,6+,8+;;/m0../s1

InChI key

ABWVCNMFYVEBIB-CDNBRZBRSA-L

General description

2′-Deoxycytidine 5′-triphosphate (dCTP) is synthesized either by de novo pathway or from the multiple phosphorylation steps of cytidine. It is deaminated to deoxyuridine monophosphate (dUMP) which is convertible to deoxythymidine triphosphate (dTTP).

Application

2′-Deoxycytidine 5′-triphosphate disodium salt has been used:
  • in polymerase chain reaction mix for the amplification of Alternaria (AM) toxin DNA[1]
  • to spike oligonucleotide sample for capillary electrophoresis (CE) analysis[2]
  • as a component in adenosine 5′-monophosphate (AMP) luciferase assay[3]

2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.

Biochem/physiol Actions

2′-Deoxycytidine 5′-triphosphate (dCTP) is used to synthesize DNA by DNA polymerases or reverse transcriptases. dCTP is an allosteric regulator of deoxycytidylate (dCMP) deaminase.[4] It is used in studies of nucleotide pools, nucleotide selectivity and enzyme specificity studies, such as the preferred incorporation of dCTP by Rev1, a γ-family DNA polymerase.[5]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D4913DCTP-ROD5038
Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200

form

powder

form

liquid

form

solution

form

liquid

assay

≥95%

assay

≥99%

assay

99% (dNTP)

assay

≥99%

shipped in

dry ice

shipped in

dry ice

shipped in

-

shipped in

dry ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Xiaoli Xiang et al.
Analytical biochemistry, 460, 22-28 (2014-05-27)
A luciferase-based method was developed for measurement of 5'-adenylylsulfate (APS) reductase (APR), an enzyme of the reductive sulfate assimilation pathway in prokaryotes and plants. APR catalyzes the two-electron reduction of APS and forms sulfite and adenosine 5'-monophospahate (AMP). The luciferase-based
Genetics: Analysis of Genes and Genomes (2009)
Dynamics of pyrimidine deoxynucleoside triphosphate pools in relationship to DNA synthesis in 3T6 mouse fibroblasts
Nicander B and Reichard P
Proceedings of the National Academy of Sciences, 80(5), 1347-1351 (1983)



Global Trade Item Number

SKUGTIN
D4635-1G04061826690567
D4635-25MG04061833006887
D4635-100MG04061833565575
D4635-10MG04061833006870

Questions

Reviews

No rating value

Active Filters