Skip to Content
Merck
All Photos(1)

Key Documents

SML1220

Sigma-Aldrich

Doripenem hydrate

≥98% (HPLC)

Synonym(s):

(4R,5S,6S)-3-[[(3S,5S)-5-[[(Aminosulfonyl)amino]methyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N4O6S2 · H2O
CAS Number:
Molecular Weight:
438.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC[C@H]1NC[C@H](C1)SC2=C(N3[C@H]([C@H]2C)[C@H](C3=O)[C@H](O)C)C(=O)O)N.O

InChI

1S/C15H24N4O6S2.H2O/c1-6-11-10(7(2)20)14(21)19(11)12(15(22)23)13(6)26-9-3-8(17-5-9)4-18-27(16,24)25;/h6-11,17-18,20H,3-5H2,1-2H3,(H,22,23)(H2,16,24,25);1H2/t6-,7-,8+,9+,10-,11-;/m1./s1

InChI key

NTUBEBXBDGKBTJ-WGLOMNHJSA-N

Biochem/physiol Actions

Doripenem hydrate is used to treat complicated urinary infection including Pyelonephritis caused by E.coli. Doripenem hydrate is promoted in the United States as DORIBAX®. This drug is synthesized from p-nitrobenzyl-protected enolphosphate 2b and N-(p-nitrobenzyloxycarbonyl)-protected aminomethylpyrrolidine.
Doripenem is an ultra-broad spectrum carbopenem antibiotic.
Doripenem is an ultra-broad spectrum carbopenem antibiotic. Doripenem is active against both gram-positive and gram-negative bacteria.

Legal Information

DORIBAX is a registered trademark of Shionogi Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical large-scale synthesis of doripenem: A novel 1β-methylcarbapenem antibiotic.
Nishino Y, et al.
Organic Process Research & Development, 7(6), 846-850 (2003)
Improved Large-scale One-pot Synthesis of Pure Doripenem Hydrate (S-4661).
Raju K R, et al.
Organic Prep. and Proc. Int., 48(4), 342-349 (2016)
Lokender Kumar et al.
Frontiers in microbiology, 12, 598498-598498 (2021-02-16)
Pseudomonas aeruginosa utilizes the quorum sensing (QS) system to strategically coordinate virulence and biofilm formation. Targeting QS pathways may be a potential anti-infective approach to treat P. aeruginosa infections. In the present study, we define cephalosporins' anti-QS activity using Chromobacterium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service