Skip to Content
Merck
All Photos(1)

Key Documents

SML3869

Sigma-Aldrich

Avibactam Sodium

≥98% (HPLC)

Synonym(s):

(2S,5R)-7-Oxo-6-(sulfooxy)-1,6-diazabicyclo[3.2.1]octane-2-carboxamide monosodium salt, AVE1330A sodium salt, NXL-104 sodium salt, Sulfuric acid mono[(1R,2S,5R)-2-(aminocarbonyl)-7-oxo-1,6-diazabicyclo[3.2.1]oct-6-yl] ester sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N3NaO6S
CAS Number:
Molecular Weight:
287.23
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[Na+].[S](=O)(=O)([O-])ON1[C@H]2CN([C@@H](CC2)C(=O)N)C1=O

InChI key

RTCIKUMODPANKX-JBUOLDKXSA-M

Biochem/physiol Actions

Avibactam is a reversible non-β-lactam β-lactamase inhibitor with IC50 values of 5 nM for CTX-M-15, 8nM for TEM-1, and 80 nM for P99 β-lactamase enzymes.
Reversible non-β-lactam, β-lactamase inhibitor

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbapenemase Inhibitors: Updates on Developments in 2021
Journal of clinical medicine research, 14(7), 251-259 (2022)
Efficacies of ceftazidime-avibactam and ceftazidime against Pseudomonas aeruginosa in a murine lung infection model
Antimicrobial Agents and Chemotherapy, 58(3), 1365-1371 (2014)
Avibactam is a covalent, reversible, non-?-lactam ?-lactamase inhibitor
Proceedings of the National Academy of Sciences of the USA, 109(29), 11663-11668 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service