Skip to Content
Merck
All Photos(1)

Key Documents

A2487

Sigma-Aldrich

Cotrimoxazole, Ready Made Solution

100 mg/mL in DMSO

Synonym(s):

Trimethoprim-Sulfamethoxazole, TMP-SMX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O3 · 5 C10H11N3O3S
CAS Number:
Molecular Weight:
1556.71
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85

form

liquid

Quality Level

concentration

100 mg/mL in DMSO

color

colorless to faint yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
parasites

Mode of action

DNA synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

[S](=O)(=O)(Nc4n[o]c(c4)C)c3ccc(cc3)N.Nc1nc(c(cn1)Cc2cc(c(c(c2)OC)OC)OC)N

InChI

1S/C14H18N4O3.C10H11N3O3S/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15;1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h5-7H,4H2,1-3H3,(H4,15,16,17,18);2-6H,11H2,1H3,(H,12,13)

InChI key

WZRJTRPJURQBRM-UHFFFAOYSA-N

Application

Cotrimoxazole has been used as an antimicrobial agent.

Biochem/physiol Actions

Cotrimoxazole, a combination of trimethoprim and sulfamethoxazole, interferes with the cellular metabolism of folic acid in the bacterial cell by blocking the biosynthesis of nucleotides. Trimethoprim binds to dihydrofolate reductase (DHFR) which inhibits the reduction of dihydrofolic acid (DHF) to tetrahydrofolic acid (THF), resulting in an antimicrobial effect. Sulfamethoxazole interferes with the synthesis of nucleic acids in sensitive microorganisms by blocking the conversion of p-aminobenzoic acid to the coenzyme dihydrofolic acid. The net effect of these actions is to inhibit thymidine synthesis which prevents bacterial DNA synthesis.

Packaging

1ml

Preparation Note

prepared using a 1:5 mixture of trimethoprim:sulfamethoxazole

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diversity of Escherichia coli strains producing extended-spectrum beta-lactamases in Spain: second nationwide study.
Diaz MA, et al.
Journal of Clinical Microbiology, 48, 2840-2845 (2010)
Escherichia coli resistant to quinolones in a neonatal unit.
Ruiz Del Castillo B, et al.
Journal of Medical Microbiology, 60, 1713-1716 (2011)
Amy J Pate et al.
The American journal of emergency medicine, 32(2), 135-138 (2013-11-19)
Methicillin-resistant Staphylococcus aureus (MRSA) infections are becoming increasingly prevalent in both community and hospital settings. Certain strains are notorious for causing skin and soft tissue infections in patients with no established risk factors. In this article, we report our findings
Lise Denoeud-Ndam et al.
Journal of acquired immune deficiency syndromes (1999), 65(2), 198-206 (2013-11-14)
Malaria during pregnancy has serious consequences that are worsened by HIV infection. Malaria preventive measures for HIV-infected pregnant women include cotrimoxazole (CTX) prophylaxis given to prevent HIV-related opportunistic infections and also protective against malaria, or intermittent preventive treatment (IPTp) with
H Abolhassani et al.
Journal of investigational allergology & clinical immunology, 23(5), 323-329 (2013-11-23)
Autoimmune disorders occur with a higher incidence in common variable immunodeficiency (CVID) patients than in the general population. To describe the clinical features of the autoimmune phenotype in patients with CVID. The hospital records of all diagnosed CVID patients referred

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service