Skip to Content
Merck
All Photos(1)

Key Documents

SML1238

Sigma-Aldrich

Ertapenem sodium

≥90% (HPLC), powder, carbopenem antibiotic

Synonym(s):

(4R,5S,6S)-3-[(3S,5S)-5-[(3-carboxyphenyl)carbamoyl]pyrrolidin-3-yl]sulfanyl-6-(1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid sodium, ZD-443 sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N3O7S · Na
CAS Number:
Molecular Weight:
497.50
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Ertapenem sodium, ≥90% (HPLC)

Quality Level

Assay

≥90% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

H2O: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−70°C

SMILES string

[Na+].S([C@@H]3CN[C@@H](C3)C(=O)Nc4cc(ccc4)C(=O)[O-])C1=C(N2[C@H]([C@H]1C)[C@H](C2=O)[C@H](O)C)C(=O)O

InChI

1S/C22H25N3O7S.Na/c1-9-16-15(10(2)26)20(28)25(16)17(22(31)32)18(9)33-13-7-14(23-8-13)19(27)24-12-5-3-4-11(6-12)21(29)30;/h3-6,9-10,13-16,23,26H,7-8H2,1-2H3,(H,24,27)(H,29,30)(H,31,32);/q;+1/p-1/t9-,10-,13+,14+,15-,16-;/m1./s1

InChI key

ZXNAQFZBWUNWJM-HRXMHBOMSA-M

Application

Ertapenem sodium has been used as a carbapenem in the NitroSpeed-Carba NP test in minimum inhibitory concentration (MIC) testing for differentiation of carbapenemases in Enterobacterales and Pseudomonas aeruginosa. It has also been used as an antibiotic to study its effects on the survival of septic mice.

Biochem/physiol Actions

Broad spectrum carbopenem antibiotic
Ertapenem is a broad spectrum carbopenem antibiotic. Ertapenem is active against both gram-positive and gram-negative bacteria. Ertapenem shows an extended duration of action in vivo due to its reversible binding to serum protein.
Ertapenem sodium exhibits its activity against several aerobic and anaerobic microorganisms and is more resistant to β-lactamase enzymes. It has been studied in the treatment of several infectious diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas J MacVittie et al.
Health physics, 109(5), 427-439 (2015-10-02)
A nonhuman primate (NHP) model of acute high-dose, partial-body irradiation with 5% bone marrow (PBI/BM5) sparing was used to assess the effect of Neupogen® [granulocyte colony stimulating factor (G-CSF)] to mitigate the associated myelosuppression when administered at an increasing interval
Tahisa Marcela Pedroso et al.
Critical reviews in analytical chemistry, 46(1), 15-21 (2016-02-10)
Antimicrobials have unquestionable importance in the control of many diseases; however the constant concern with evolution of resistant microorganisms is increasing. Ertapenem sodium is a β-lactam antibiotic of the carbapenem class that has a broader activity spectrum than most other
Daniele C Nascimento et al.
Nature communications, 8, 14919-14919 (2017-04-05)
Patients who survive sepsis can develop long-term immune dysfunction, with expansion of the regulatory T (Treg) cell population. However, how Treg cells proliferate in these patients is not clear. Here we show that IL-33 has a major function in the
Raphael G Ferreira et al.
The Journal of infection, 77(5), 391-397 (2018-09-19)
Sepsis is an overwhelming systemic inflammation resulting from an uncontrolled infection that causes extensive tissue damage, organ dysfunction and eventually death. A growing body of evidence indicates that impaired neutrophil migration to the site of infection is associated with poor
Lingbo Wang et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 106, 415-420 (2021-04-18)
This study aimed to evaluate the performance of the NitroSpeed-Carba NP test for detecting carbapenemases in the clinical strains of Enterobacterales and Pseudomonas aeruginosa (P. aeruginosa), and analyze its advantages and limitations. The antimicrobial susceptibility tests were performed according to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service