Skip to Content
Merck
All Photos(1)

Key Documents

SML1134

Sigma-Aldrich

I-CBP112

≥98% (HPLC)

Synonym(s):

1-[7-(3,4-Dimethoxyphenyl)-9-[[(3S)-1-methylpiperidin-3-yl]methoxy]-2,3,4,5-tetrahydr­o-1,4-benzoxazepin-4-yl]propan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36N2O5
CAS Number:
Molecular Weight:
468.59
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1)CCC[C@@H]1COC2=CC(C3=CC=C(OC)C(OC)=C3)=CC4=C2OCCN(C(CC)=O)C4

InChI

1S/C27H36N2O5/c1-5-26(30)29-11-12-33-27-22(17-29)13-21(20-8-9-23(31-3)24(14-20)32-4)15-25(27)34-18-19-7-6-10-28(2)16-19/h8-9,13-15,19H,5-7,10-12,16-18H2,1-4H3/t19-/m0/s1

InChI key

YKNAKDFZAWQEEO-IBGZPJMESA-N

Application

I-CBP112 has been used in cell culture and sample preparation, to verify the acetylation of VPS34 (vacuolar protein sorting 34), by p300 at K29, K771 and K781 and to diminish synergistic gene expression.

Biochem/physiol Actions

I-CBP112 is a selective inhibitor of the bromodomain-containing transcription factors CREBBP (CBP) and EP300 (IC50 = 0.142 and 0.625 μM, respectively). I-CBP112 has little activity against other bromodomains at concentrations up to 1 mM. For full characterization details, please see I-CBP112 on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
I-CBP112 is a selective inhibitor of the bromodomain-containing transcription factors CREBBP (CBP) and EP300.
I-CBP112 is also known as (1-[7-(3,4-dimethoxyphenyl)-9-{[(3S)-1-methylpiperidin-3-yl]-methoxy}-2,3,4,5-tetrahydro-1,4-benzoxazepin-4-yl]propan-1-one). It has the ability to induce full-length p300/CBP nucleosome acetylation. It is a competitive inhibitor of acetyl-lysine. I-CBP112 regulates transcription of key inflammatory genes.

Features and Benefits

I-CBP112 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

I-CBP112 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the I-CBP112 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of a selective small molecule inhibitor of the CBP/p300 bromodomain for leukemia therapy.
Picaud S, et al.
Cancer Research, 75(23), 5106-5119 (2015)
VPS34 Acetylation Controls Its Lipid Kinase Activity and the Initiation of Canonical and Non-canonical Autophagy.
Su H, et al.
Molecular Cell, 67(6), 907-921 (2017)
Modulation of p300/CBP Acetylation of Nucleosomes by Bromodomain Ligand I-CBP112.
Zucconi BE, et al.
Biochemistry, 55(27), 3727-3734 (2016)
Time-Resolved Analysis Reveals Rapid Dynamics and Broad Scope of the CBP/p300 Acetylome.
Weinert BT, et al.
Cell, 174(1), 231-244 (2018)
Synergistic gene expression during the acute phase response is characterized by transcription factor assisted loading.
Goldstein I, et al.
Nature Communications, 8(1), 1849-1849 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service