Skip to Content
Merck
All Photos(1)

Documents

P13207

Sigma-Aldrich

Phenazine

98%

Synonym(s):

Dibenzo[b,e]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2
CAS Number:
Molecular Weight:
180.21
Beilstein:
126500
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

crystals

mp

172-176 °C (lit.)

fluorescence

λex 426 nm; λem 471 nm in ethyl acetate

SMILES string

c1ccc2nc3ccccc3nc2c1

InChI

1S/C12H8N2/c1-2-6-10-9(5-1)13-11-7-3-4-8-12(11)14-10/h1-8H

InChI key

PCNDJXKNXGMECE-UHFFFAOYSA-N

Gene Information

human ... PRNP(5621)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Wang et al.
Science (New York, N.Y.), 364(6446) (2019-06-22)
Ruminants are the only extant mammalian group possessing bony (osseous) headgear. We obtained 221 transcriptomes from bovids and cervids and sequenced three genomes representing the only two pecoran lineages that convergently lack headgear. Comparative analyses reveal that bovid horns and
Daniel W Watkins et al.
Nature communications, 8(1), 358-358 (2017-08-27)
Although catalytic mechanisms in natural enzymes are well understood, achieving the diverse palette of reaction chemistries in re-engineered native proteins has proved challenging. Wholesale modification of natural enzymes is potentially compromised by their intrinsic complexity, which often obscures the underlying
Steven Higgins et al.
Frontiers in cellular and infection microbiology, 8, 252-252 (2018-08-08)
The Pseudomonas aeruginosa quorum sensing (QS) network plays a key role in the adaptation to environmental changes and the control of virulence factor production in this opportunistic human pathogen. Three interlinked QS systems, namely las, rhl, and pqs, are central
Wei-Liang Kong et al.
Microorganisms, 8(4) (2020-04-25)
Plant growth-promoting rhizobacteria (PGPR) can potentially be used as an alternative strategy to control plant diseases. In this study, strain ST-TJ4 isolated from the rhizosphere soil of a healthy poplar was found to have a strong antifungal activity against 11
Biancastella Cereser et al.
The Journal of pathology, 244(1), 61-70 (2017-09-25)
It is widely accepted that the cell of origin of breast cancer is the adult mammary epithelial stem cell; however, demonstrating the presence and location of tissue stem cells in the human breast has proved difficult. Furthermore, we do not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service