Skip to Content
Merck
All Photos(1)

Documents

SML1037

Sigma-Aldrich

UNC0642

≥98% (HPLC)

Synonym(s):

2-(4,4-Difluoropiperidin-1-yl)-6-methoxy-N-[1-(propan-2-yl)piperidin-4-yl]-7-[3-(pyrrolidin-1-yl)propoxy]quinazolin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H44F2N6O2
CAS Number:
Molecular Weight:
546.70
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC(CC1)(F)CCN1C2=NC(NC3CCN(C(C)C)CC3)=C4C(C=C(OCCCN5CCCC5)C(OC)=C4)=N2

InChI

1S/C29H44F2N6O2/c1-21(2)36-14-7-22(8-15-36)32-27-23-19-25(38-3)26(39-18-6-13-35-11-4-5-12-35)20-24(23)33-28(34-27)37-16-9-29(30,31)10-17-37/h19-22H,4-18H2,1-3H3,(H,32,33,34)

InChI key

RNAMYOYQYRYFQY-UHFFFAOYSA-N

Application

UNC0642 has been used to treat human HeLa cells to inhibit the DNA ligase 1 (LIG1) /UHRF1 (ubiquitin-like with PHD and ring finger domains 1) interaction.

Biochem/physiol Actions

UNC0642 is a potent, selective inhibitor of histone methyltransferases G9a (EHMT2) and GLP (EHMT1), which catalyze the mono and dimethylation of lysine 9 of histone 3 (H3K9), and other non-histone substrates such as p53 and WIZ. UNC0642 has an in vitro IC50 <15 nM with greater than 100-fold selectivity over 13 other HMTs and selected representatives of kinases, ion channels, 7TMs, and other epigenetic proteins. UNC0642 has the same potency with improved PK properties relative to UNC0638, which should make it a more useful probe in an in vivo setting. For full characterization details, please visit the UNC0642 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
UNC0642 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

Other Notes

UNC0642 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the UNC0642 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiffani D M Berkel et al.
The international journal of neuropsychopharmacology, 22(4), 292-302 (2018-12-28)
Tolerance to ethanol-induced anxiolysis promotes alcohol intake, thus contributing to alcohol use disorder development. Recent studies implicate histone deacetylase-mediated histone H3K9 deacetylation in regulating neuropeptide Y expression during rapid ethanol tolerance to the anxiolytic effects of ethanol. Furthermore, the histone
Yue-Peng Cao et al.
Acta pharmacologica Sinica, 40(8), 1076-1084 (2019-02-16)
Urinary bladder cancer (UBC) is characterized by frequent recurrence and metastasis despite the standard chemotherapy with gemcitabine and cisplatin combination. Histone modifiers are often dysregulated in cancer development, thus they can serve as an excellent drug targets for cancer therapy.
Patricia D B Tiburcio et al.
Translational oncology, 13(10), 100819-100819 (2020-07-06)
Malignant gliomas have disproportionally high morbidity and mortality. Heterozygous mutations in the isocitrate dehydrogenase 1 (IDH1) gene are most common in glioma, resulting in predominantly arginine to histidine substitution at codon 132. Because IDH1R132H requires a wild-type allele to produce
Methylation of DNA Ligase 1 by G9a/GLP Recruits UHRF1 to Replicating DNA and Regulates DNA Methylation.
Ferry L, et al.
Molecular Cell, 67(4), 550-565 (2017)
Chengfa Zhao et al.
Reproduction (Cambridge, England), 157(4), 359-369 (2019-02-08)
Somatic cell nuclear transfer in mammalian cloning suffers from a faulty epigenetic reprogramming, which is believed to cause developmental failures in cloned embryos. Regulating the epigenetic-modifying enzymes can rescue the chromatin of cloned embryos from aberrant epigenetic status, thereby potentially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service