Skip to Content
Merck
All Photos(1)

Key Documents

SML0352

Sigma-Aldrich

PFI-1

≥98% (HPLC)

Synonym(s):

2-Methoxy-N-(3-methyl-2-oxo-1,2,3,4-tetrahydro-quinazolin-6-yl)-bezenesulfonamide, PF-06405761

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O4S
CAS Number:
Molecular Weight:
347.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccccc1S(=O)(=O)Nc2ccc3NC(=O)N(C)Cc3c2

InChI

1S/C16H17N3O4S/c1-19-10-11-9-12(7-8-13(11)17-16(19)20)18-24(21,22)15-6-4-3-5-14(15)23-2/h3-9,18H,10H2,1-2H3,(H,17,20)

InChI key

TXZPMHLMPKIUGK-UHFFFAOYSA-N

Application

PFI-1 has been used as a bromodomain inhibitor in breast cancer cell line MCF7 dechorionated zebrafish embryos. It has also been used in human peripheral blood mononuclear cells (PBMCs), to inhibit the release of interleukin-6 in lipopolysaccharide (LPS) challenge assay.

Biochem/physiol Actions

PFI-1 is a quinazolinone inhibits androgen receptor (AR) mediated signaling in prostate cancer cells. It elicits minimum toxicity and is effective for anti-HIV-latency therapy. In leukemia cells, PFI-1 inhibits Aurora B kinase and promotes caspase-dependent apoptosis.
Selective BET (bromodomain-containing protein) inhibitor.
The human BET family, which includes BRD2, BRD3, BRD4 and BRDT, play a role in regulation of gene transcription. PFI-1 is a selective BET bromodomain inhibitor with activity at BRD2 (IC50 = 98 nM) and BRD4 (IC50 = 220 nM). For full characterization details, please visit the PFI-1 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

PFI-1 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

PFI-1 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the PFI-1 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Estrogen-dependent control and cell-to-cell variability of transcriptional bursting
Fritzsch C, et al.
Molecular Systems Biology, 14(2) (2018)
Marie C Hupe et al.
World journal of urology, 37(2), 343-349 (2018-06-24)
The bromodomain and extra-terminal (BET) family of proteins provides a scaffolding platform for the recruitment and tethering of transcription factors to acetylated chromatin, thereby modulating gene expression. In this study, we evaluated the efficacy of the BET-inhibitor PFI-1 to diminish
BET inhibitors RVX-208 and PFI-1 reactivate HIV-1 from latency
Lu P, et al.
Scientific reports, 7(1), 16646-16646 (2017)
Justin Hall et al.
Biochemical and biophysical research communications, 527(1), 250-256 (2020-05-25)
DNA-encoded libraries (DELs) can contain billions of unique chemical species; selecting against such large inputs, it is typical to find more candidate binders than is reasonable to pursue for follow-up synthesis and testing. Given this wealth of choices, common practice
Design and chemoproteomic functional characterization of a chemical probe targeted to bromodomains of BET family proteins
Wu J, et al.
MedChemComm, 5(12), 1871-1878 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service