Skip to Content
Merck
All Photos(1)

Key Documents

SML0936

Sigma-Aldrich

Teriflunomide

≥98% (HPLC)

Synonym(s):

(Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide, A 1726, A77-1726, A771726, Flucyamide, HMR 1726, N-(4-Trifluoromethylphenyl)-2-cyano-3-hydroxycrotoamide, SU 20

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9F3N2O2
CAS Number:
Molecular Weight:
270.21
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C12H9F3N2O2/c1-7(18)10(6-16)11(19)17-9-4-2-8(3-5-9)12(13,14)15/h2-5,18H,1H3,(H,17,19)/b10-7-

InChI key

UTNUDOFZCWSZMS-YFHOEESVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Teriflunomide has been used as a dihydroorotate dehydrogenase (DHODH) inhibitor.

Biochem/physiol Actions

Teriflunomide is an orally available anti-inflammatory immunomodulator.
Teriflunomide is an orally available anti-inflammatory immunomodulator. It blocks the activity of dihydroorotate dehydrogenase, preventing pyrimidine synthesis and T and B cell proliferation and function. Teriflunomide has been used to treat rheumatoid arthritis and was recently approved for multiple sclerosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of dihydroorotate dehydrogenase overcomes differentiation blockade in acute myeloid leukemia.
Sykes D B, et al.
Cell, 167(1), 171-186 (2016)
Mieradilijiang Abudupataer et al.
eLife, 10 (2021-09-07)
Bicuspid aortic valve (BAV) is the most common congenital cardiovascular disease in general population and is frequently associated with the development of thoracic aortic aneurysm (TAA). There is no effective strategy to intervene with TAA progression due to an incomplete
Zhen-Guo Ma et al.
Clinical science (London, England : 1979), 132(6), 685-699 (2018-03-16)
T-cell infiltration and the subsequent increased intracardial chronic inflammation play crucial roles in the development of cardiac hypertrophy and heart failure (HF). A77 1726, the active metabolite of leflunomide, has been reported to have powerful anti-inflammatory and T cell-inhibiting properties.
Andrea Bilger et al.
Oncotarget, 8(27), 44266-44280 (2017-06-03)
EBV infection causes mononucleosis and is associated with specific subsets of B cell lymphomas. Immunosuppressed patients such as organ transplant recipients are particularly susceptible to EBV-induced lymphoproliferative disease (LPD), which can be fatal. Leflunomide (a drug used to treat rheumatoid
Deepti Mathur et al.
Cancer discovery, 7(4), 380-390 (2017-03-04)
Metabolic changes induced by oncogenic drivers of cancer contribute to tumor growth and are attractive targets for cancer treatment. Here, we found that increased growth of PTEN-mutant cells was dependent on glutamine flux through the de novo pyrimidine synthesis pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service