Skip to Content
Merck
All Photos(1)

Key Documents

C5874

Sigma-Aldrich

Cidofovir hydrate

≥98% (HPLC), powder, viral DNA synthesis inhibitor

Synonym(s):

(S)-1-[3-hydroxy-2-(phosphonylmethoxy)propyl]cytosine, (S)-HPMPC; Cidovir, HPMPC, Vistide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N3O6P · xH2O
CAS Number:
Molecular Weight:
279.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Cidofovir hydrate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

solubility

deionized water: ≥5 mg/mL (warmed)

storage temp.

room temp

SMILES string

O.NC1=NC(=O)N(C[C@@H](CO)OCP(O)(O)=O)C=C1

InChI

1S/C8H14N3O6P.H2O/c9-7-1-2-11(8(13)10-7)3-6(4-12)17-5-18(14,15)16;/h1-2,6,12H,3-5H2,(H2,9,10,13)(H2,14,15,16);1H2/t6-;/m0./s1

InChI key

KLJYJZIIJLYJNN-RGMNGODLSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective inhibitor of viral DNA synthesis through the selective inhibition of viral DNA polymerase.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen D S McCarthy et al.
PLoS neglected tropical diseases, 10(1), e0004364-e0004364 (2016-01-12)
To date there are no approved antiviral drugs for the treatment of Ebola virus disease (EVD). While a number of candidate drugs have shown limited efficacy in vitro and/or in non-human primate studies, differences in experimental methodologies make it difficult
Michel Bossens et al.
International journal of clinical pharmacology and therapeutics, 56(3), 134-141 (2018-01-11)
The primary objective was to evaluate the safety and local tolerance of a topical 2% (w/w) cidofovir gel, applied directly to the cervices of women with high-grade cervical intraepithelial neoplasia (CIN 2+). The secondary objective was to evaluate the pharmacokinetics
Erika Trompet et al.
Antiviral research, 182, 104901-104901 (2020-08-09)
Murine γ-herpesvirus-68 (MHV-68), genetically and biologically related to human γ-herpesviruses Epstein-Barr virus and Kaposi's sarcoma-associated herpesvirus, can be easily propagated in vitro allowing drug resistance studies. Previously, we described specific changes in MHV-68 protein kinase (PK) or thymidine kinase (TK)
Joanna Jackowska et al.
Lasers in medical science, 34(7), 1433-1440 (2019-02-15)
Mechanical debulking of laryngeal papillomas is associated with voice disorders due to impairment of the vocal fold's mucosa. Intraepithelial injection of cidofovir reduces damage to the laryngeal structures and thus improves vocal parameters. The aim of our study was to
Successful treatment of perianal giant condyloma acuminatum in an immunocompromised host with systemic interleukin 2 and topical cidofovir.
Vinod E Nambudiri et al.
JAMA dermatology, 149(9), 1068-1070 (2013-07-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service