Skip to Content
Merck
All Photos(1)

Key Documents

SML0040

Sigma-Aldrich

Lometrexol hydrate

≥95% (HPLC), powder, GARFTase inhibitor

Synonym(s):

LY 264618 hydrate, N-[4-[2-[(6R)-2-amino-3,4,5,6,7,8-hexahydro-4-oxopyrido[2,3-d]pyrimidin-6-yl]ethyl]benzoyl]-L-glutamic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N5O6 · xH2O
CAS Number:
Molecular Weight:
443.45 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Lometrexol hydrate, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to light yellow

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

O.NC1=NC(=O)C2=C(NC[C@H](CCc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)C2)N1

InChI

1S/C21H25N5O6.H2O/c22-21-25-17-14(19(30)26-21)9-12(10-23-17)2-1-11-3-5-13(6-4-11)18(29)24-15(20(31)32)7-8-16(27)28;/h3-6,12,15H,1-2,7-10H2,(H,24,29)(H,27,28)(H,31,32)(H4,22,23,25,26,30);1H2/t12-,15+;/m1./s1

InChI key

AEFQSKJUVDZANQ-YLCXCWDSSA-N

Application

Lometrexol hydrate was used to compare the biological activity of potent inhibitor of human GARFTase.

Biochem/physiol Actions

Glycinamide Ribonucleotide Formyltransferase (GARFTase) is a folate-dependent enzyme required for de novo purine synthesis. Lometrexate is a potent inhibitor of GARFTase, but does not interfere with enzymes involved in the synthesis of folate. Lometrexerol has been tested clinically for the treatment of various cancers as an anti-folate like agent, similar to methotrexate. Treatment with lometrexol rapidly decreases ATP and GTP levels, cell cycle arrest and induces apoptosis. Although depletion of nucleotide pools induces p53 expression, lometrexol is cytotoxic in both wild-type and mutant p53 expressing tumor cells. Lometrexol is cytotoxic in CCRF-CEm leukemia cells with an IC50 of 2.9 nM.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New agents in cancer clinical trials.
J Adams et al.
Oncogene, 19(56), 6687-6692 (2001-06-28)
D R Newell
Seminars in oncology, 26(2 Suppl 6), 74-81 (1999-12-22)
Antifolate drugs, as a class, have broad-spectrum activity against both hematologic and solid human malignancies. The pharmacokinetics of the classical antifolate methotrexate have been well-defined and pharmacokinetic data can be exploited to reduce the toxicity and enhance the activity of
X Lu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 7(7), 2114-2123 (2001-07-13)
The impact of p53 status on cellular sensitivity to antifolate drugs has been examined in seven human cell lines (A549, MCF7, T-47D, CCRF-CEM, COR-L23, A2780, and HCT-116) and p53 nonfunctional counterparts of two of the cell lines (HCT-116/N7 and A2780/CP70).
C Shih et al.
Advances in enzyme regulation, 38, 135-152 (1998-10-08)
Extensive biochemical and pharmacological evidence indicates that LY231514 is a novel antifolate antimetabolite. LY231514 is transported into cells mainly through the reduced folate carrier system and extensively metabolized to polyglutamated forms. The polyglutamates of LY231514 inhibit at least three key
Huiling Qi et al.
Cancer research, 66(11), 5875-5882 (2006-06-03)
The folate receptor (FR) type beta is a promising target for therapeutic intervention in acute myelogenous leukemia (AML), owing particularly to its selective up-regulation in the leukemic cells by all-trans retinoic acid (ATRA). Here we show, using KG-1 and MV4-11

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service