Skip to Content
Merck
All Photos(1)

Key Documents

SML0491

Sigma-Aldrich

Ritonavir

≥98% (HPLC)

Synonym(s):

A-84538, ABT-538, Abbott 84538

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48N6O5S2
CAS Number:
Molecular Weight:
720.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution, warmed)

storage temp.

room temp

SMILES string

O=C(OCC1=CN=CS1)N[C@H]([C@@H](O)C[C@@H](NC([C@@H](NC(N(CC2=CSC(C(C)C)=N2)C)=O)C(C)C)=O)CC3=CC=CC=C3)CC4=CC=CC=C4

InChI

1S/C37H48N6O5S2/c1-24(2)33(42-36(46)43(5)20-29-22-49-35(40-29)25(3)4)34(45)39-28(16-26-12-8-6-9-13-26)18-32(44)31(17-27-14-10-7-11-15-27)41-37(47)48-21-30-19-38-23-50-30/h6-15,19,22-25,28,31-33,44H,16-18,20-21H2,1-5H3,(H,39,45)(H,41,47)(H,42,46)/t28-,31-,32-,33-/m0/s1

InChI key

NCDNCNXCDXHOMX-XGKFQTDJSA-N

Application

Ritonavir has been used:
  • as a human immunodeficiency virus (HIV) protease inhibitor to study its effects on placental endocrine function
  • as an HIV protease inhibitor to study its effects on reduction of tetrazolium dye in human embryonic kidney cells
  • as an organic anion transporting polypeptide (OATP) inhibitor to study its effect on hepatic uptake of bergapten and imperatorin

Biochem/physiol Actions

Ritonavir is an HIV protease inhibitor now used frequently as a booster of other protease inhbitors. Ritonavir inhibits cytochrome P450-3A4 (CYP3A4), a liver enzyme that normally metabolizes protease inhibitors. It has also been investigated as a possible anti-cancer agent.
Ritonavir is an HIV protease inhibitor; antiretroviral.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José R Santos et al.
The Journal of antimicrobial chemotherapy, 70(4), 1124-1129 (2014-12-20)
Data on the efficacy of simplifying therapy using darunavir/ritonavir and lopinavir/ritonavir monotherapy in clinical practice remain limited. A retrospective single-centre study including patients initiating darunavir/ritonavir or lopinavir/ritonavir monotherapy with a plasma HIV-1 viral load (pVL) <50 copies/mL and at least
Lin Chen et al.
Journal of ethnopharmacology, 212, 74-85 (2017-10-23)
Radix Angelica dahuricae (RAD), the roots of Angelica dahurica (Hoffm.) Benth. & Hook.f. ex Franch. & Sav, is a well-known traditional Chinese medicine (TCM) and has been used for centuries to treat headaches, toothaches, nose congestion, abscesses, furunculoses, and acne.
Mary S Ngoma et al.
Journal of the International AIDS Society, 18, 19352-19352 (2015-07-04)
To prevent mother-to-child transmission (MTCT) of HIV in developing countries, new World Health Organization (WHO) guidelines recommend maternal combination antiretroviral therapy (cART) during pregnancy, throughout breastfeeding for 1 year and then cessation of breastfeeding (COB). The efficacy of this approach
Laura Ciaffi et al.
AIDS (London, England), 29(12), 1473-1481 (2015-08-06)
WHO recommends ritonavir-boosted protease inhibitor with two nucleoside reverse transcriptase inhibitors in HIV-infected patients failing non-nucleoside reverse transcriptase inhibitor-based first-line treatment. Here, we aimed to provide more evidence for the choice of nucleoside reverse transcriptase inhibitor and boosted protease inhibitor.
Maria Casadellà et al.
AIDS (London, England), 29(12), 1493-1504 (2015-08-06)
This article aims to investigate if the detection of preexisting drug-resistant minority variant (DRMV) and/or X4 HIV-1 variants could improve the efficacy of first-line combined antiretroviral therapy (ART) in late presenters. Post-hoc, combined analysis of two open-label, prospective, randomized clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service