Skip to Content
Merck
All Photos(1)

Documents

SML0309

Sigma-Aldrich

Sodium phenylbutyrate

≥98% (HPLC)

Synonym(s):

4-PBA, 4-Phenylbutyric acid, 4-phenylbutyrate, Buphenyl, Sodium 4-phenylbutyrate, TriButyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NaO2
CAS Number:
Molecular Weight:
186.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: ≥5 mg/mL

storage temp.

room temp

SMILES string

[Na+].[O-]C(=O)CCCc1ccccc1

InChI

1S/C10H12O2.Na/c11-10(12)8-4-7-9-5-2-1-3-6-9;/h1-3,5-6H,4,7-8H2,(H,11,12);/q;+1/p-1

InChI key

VPZRWNZGLKXFOE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phenylbutyrate stimulates urinary nitrogen excretion. It is a prodrug, which undergoes β-oxidation to phenylacetate. Sodium phenylbutyrate is used to treat urea cycle disorder. It might be associated with menstrual dysfunction, amenorrhea and painful mucositis of the esophagus and stomach.

Application

Sodium phenylbutyrate has been used as an endoplasmic reticulum (ER) stress inhibitor to study the role of ER stress in the expression of betatrophin.

Biochem/physiol Actions

Sodium phenylbutyrate is a histone deacetylase inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Molecular and Genetic Basis of Neurologic and Psychiatric Disease (2008)
Phase 2 comparison of a novel ammonia scavenging agent with sodium phenylbutyrate in patients with urea cycle disorders: safety, pharmacokinetics and ammonia control
Lee B, et al.
Molecular Genetics and Metabolism, 100(3), 221-228 (2010)
Association between betatrophin/ANGPTL8 and non-alcoholic fatty liver disease: animal and human studies
Lee YH, et al.
Scientific Reports, 6(3), 24013-24013 (2016)
Xiujing Dou et al.
Food & nutrition research, 62 (2018-12-24)
Host defense peptides (HDPs) possess direct antibacterial, antineoplastic, and immunomodulatory abilities, playing a vital role in innate immunity. Dietary-regulated HDP holds immense potential as a novel pathway for preventing infection. In this study, we examined the regulation mechanism of HDPs
Advances in Clinical Chemistry, Volume 17 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service