Skip to Content
Merck
All Photos(1)

Key Documents

Z4775

Sigma-Aldrich

Zebularine

≥98% (HPLC), solid

Synonym(s):

1-(β-D-Ribofuranosyl)-1,2-dihydropyrimidin-2-one, 2-Pyrimidone-1-β-D-riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O5
CAS Number:
Molecular Weight:
228.20
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 16 mg/mL

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC=NC2=O

InChI

1S/C9H12N2O5/c12-4-5-6(13)7(14)8(16-5)11-3-1-2-10-9(11)15/h1-3,5-8,12-14H,4H2/t5-,6-,7-,8-/m1/s1

InChI key

RPQZTTQVRYEKCR-WCTZXXKLSA-N

Biochem/physiol Actions

Zebularine is a cytidine analogue, cytidine deaminase inhibitor, and DNA demethylating agent.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Li et al.
eLife, 9 (2020-05-28)
Circadian oscillations are generated via transcriptional-translational negative feedback loops. However, individual cells from fibroblast cell lines have heterogeneous rhythms, oscillating independently and with different period lengths. Here we showed that heterogeneity in circadian period is heritable and used a multi-omics
Fatima Ismail Hassan et al.
Current pharmaceutical design, 24(24), 2862-2869 (2018-09-05)
Sepsis is among the leading causes of death with no specific etiology or treatment. Increase in health burden in terms of cost, morbidity, and mortality is the reason behind the continuous search for different treatment modalities which involve several targets/approach
C B Yoo et al.
Biochemical Society transactions, 32(Pt 6), 910-912 (2004-10-28)
Regulatory genes are often hypermethylated at their promoter 5' regions and silenced in cancer. Epigenetic therapy with DNA methylation inhibitors have been shown to result in the demethylation and reactivation of these genes. Zebularine is a recently discovered mechanism-based inhibitor
Mariana Díaz et al.
The Plant cell, 31(7), 1579-1597 (2019-05-01)
The maintenance of genome integrity over cell divisions is critical for plant development and the correct transmission of genetic information to the progeny. A key factor involved in this process is the STRUCTURAL MAINTENANCE OF CHROMOSOME5 (SMC5) and SMC6 (SMC5/6)
Yukitoshi Takemura et al.
Bioscience, biotechnology, and biochemistry, 82(7), 1159-1164 (2018-04-25)
Malignant mesothelioma is an asbestos-related aggressive tumor and current therapy remains ineffective. Zebularine as a DNA methyltransferase (DNMT) inhibitor has an anti-tumor effect in several human cancer cells. The aim of the present study was to investigate whether zebularine could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service