Skip to Content
Merck
All Photos(2)

Documents

R3255

Sigma-Aldrich

13-cis-Retinoic acid

≥98% (HPLC)

Synonym(s):

Isotretinoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

yellow to yellow-orange

mp

172-175 °C (lit.)

storage temp.

−20°C

SMILES string

[H]\C(C(O)=O)=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8+,16-14-

InChI key

SHGAZHPCJJPHSC-XFYACQKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

13-cis-Retinoic acid has been used:
  • as a component of fresh media for LA-N-5 differentiation
  • to treat medulloblastoma cells for cell differentiation
  • to treat spheroids for generating time-lapse videos
  • to examine its head- deforming activity under light and dark conditions in Xenopus laevis

Biochem/physiol Actions

13-cis-Retinoic acid (RA) has anti-inflammatory and anti-tumor action. The action of RA is mediated through RAR-β and RAR-α receptors. RA attenuates iNOS expression and activity in cytokine-stimulated murine mesangial cells. It induces mitochondrial membrane permeability transition, observed as swelling and as a decrease in membrane potential, and stimulates the release of cytochrome c implicating mechanisms through the apoptosis pathway. These activities are reversed by EGTA and cyclosporin A. RA also increases MMP-1 protein expression partially via increased transcription.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Live multicellular tumor spheroid models for high-content imaging and screening in cancer drug discovery
Reid B G, et al.
Current Chemical Genomics and Translational Medicine, 8(Suppl-1), 27-27 (2014)
Induction of pronephric tubules by activin and retinoic acid in presumptive ectoderm of Xenopus laevis
Moriya N, et al.
Development, Growth & Differentiation, 35(2), 123-128 (1993)
Raoud Marayati et al.
Translational oncology, 14(1), 100893-100893 (2020-10-04)
Retinoic acid (RA) therapy has been utilized as maintenance therapy for high-risk neuroblastoma, but over half of patients treated with RA relapse. Neuroblastoma stem cell-like cancer cells (SCLCCs) are a subpopulation of cells characterized by the expression of the cell
P K Datta et al.
Kidney international, 56(2), 486-493 (1999-08-05)
Nitric oxide (NO) release as a result of cytokine-mediated activation of inducible nitric oxide synthase (iNOS) in mesangial cells can be sustained and lead to oxidative injury in various forms of glomerular inflammation. Inhibition of iNOS expression and/or activity could
M P Rigobello et al.
Biochemical pharmacology, 58(4), 665-670 (1999-07-21)
Retinoic acids, structurally related to vitamin A, inhibit the in vitro proliferation of different types of normal and neoplastic cells. The effects of all-trans, 9-cis, and 13-cis retinoic acids were tested on mitochondria isolated from rat liver. All the compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service