Skip to Content
Merck
All Photos(1)

Key Documents

SML0771

Sigma-Aldrich

LG100754

≥98% (HPLC)

Synonym(s):

(2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-2-naphthalenyl)-2,4,6-Octatrienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36O3
CAS Number:
Molecular Weight:
396.56
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O(CCC)c1cc2c(cc1\C(=C/C=C/C(=C/C(=O)O)/C)\C)C(CCC2(C)C)(C)C

InChI

1S/C26H36O3/c1-8-14-29-23-17-22-21(25(4,5)12-13-26(22,6)7)16-20(23)19(3)11-9-10-18(2)15-24(27)28/h9-11,15-17H,8,12-14H2,1-7H3,(H,27,28)/b10-9+,18-15+,19-11-

InChI key

HNODNXQAYXJFMQ-LQUSFLDPSA-N

Related Categories

Biochem/physiol Actions

LG100754 is a RXR:PPAR agonist.
LG100754 is a unique RXR ligand that acts as an antagonist against RXR homodimers, but is a strong agonist for RXR:PPARα and RXR:PPARγ heterodimers. The compound does not have agonist activities for other RXR heterodimers containing LXR or GAR/FXR receptors. LG100754 initiates adipocyte differentiation, inhibits TNFα-induced insulin receptor hypophosphorylation and improves insulin resistance in db/db mice.

Features and Benefits

This compound is featured on the InsR, Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service