Skip to Content
Merck
All Photos(5)

Key Documents

SEQXE

Sigma-Aldrich

SeqPlex DNA Amplification Kit

For use with high throughput sequencing technologies, Whole Genome Amplification kit designed to facilitate Next Gen Sequencing.

Synonym(s):

SeqPlex Enhanced DNA Amplification Kit, WGA kit, whole genome amplification kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.55

Quality Level

technique(s)

whole genome amplification: suitable

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SeqPlex Enhanced DNA Amplification Kit for whole genome amplification (WGA) is designed to facilitate next-generation sequencing (NGS) from extremely small quantities or from degraded/highly fragmented DNA. The yields from chromatin immunoprecipitation (ChIP) or formalin-fixed paraffin-embedded tissue samples (FFPE) are often less than required for successful next generation sequencing library preparation. The SeqPlex kit allows the user to pre-amplify these and other small quantity/highly fragmented DNA samples for input into a NGS workflow. This kit is an extension of the WGA product line and has been developed to integrate into the Illumina®, SOLiD, or 454 sequencing workflows.

Application

SeqPlex DNA Amplification Kit has been used for whole genome amplification.

Features and Benefits

  • Random priming technology amplifies fragmented DNA such as ChIP or FFPE
  • Facilitates sequencing from as little as 100 pg of ChIP DNA
  • Enhanced primers for complete genome coverage, minimal sequence bias, primer removal, and amplicon size ideal for next gen sequencing
  • Compatible with Illumina®, SOLiD, or 454 library prep for next generation sequencing

Other Notes

SEQXE-500RXN is manufactured on-demand. Contact technical services at [email protected] for more information.

Legal Information

Illumina is a registered trademark of Illumina, Inc.
SOLiD is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
SeqPlex is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • W4502Water, Nuclease-Free Water, for Molecular BiologySDS

  • Library Preparation Buffer

recommended

Product No.
Description
Pricing

required but not provided

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Hayakawa et al.
Cytogenetic and genome research, 161(8-9), 437-444 (2021-11-25)
E/L Repli-seq is a powerful tool for detecting cell type-specific replication landscapes in mammalian cells, but its potential to monitor DNA replication under replication stress awaits better understanding. Here, we used E/L Repli-seq to examine the temporal order of DNA
Joshua A Udall et al.
Frontiers in plant science, 10, 1541-1541 (2019-12-13)
One of the extraordinary aspects of plant genome evolution is variation in chromosome number, particularly that among closely related species. This is exemplified by the cotton genus (Gossypium) and its relatives, where most species and genera have a base chromosome
Dimiter Kunnev et al.
Journal of biological methods, 2(4) (2015-01-01)
Nascent strand capture and release (NSCR) is a method for isolation of short nascent strands to identify origins of DNA replication. The protocol provided involves isolation of total DNA, denaturation, size fractionation on a sucrose gradient, 5'-biotinylation of the appropriate
Optimizing sparse sequencing of single cells for highly multiplex copy number profiling.
Baslan T, et al.
Genome Research, 25(5), 714-724 (2015)
Isolation and sequencing of active origins of DNA replication by nascent strand capture and release (NSCR).
Kunnev D, et al.
Journal of biological methods, 2(4) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service