Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Protease Inhibitor Cocktail
Synonym(s):
Protease Inhibitor Cocktail
Products
Product Category
inhibitor cocktails (7)
inhibitors (7)
bioactive small molecules (2)
Brand
Sigma-Aldrich (16)
Biological Source
Physical Form
Tissue
Quality Segment
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
P8340
Protease Inhibitor Cocktail,
DMSO solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, for use with mammalian cell and tissue extracts, DMSO solution
Expand
View Pricing
S8830
Protease Inhibitor Cocktail,
SIGMA FAST, EDTA-Free, tablet, for the inhibition of serine, cysteine, aspartic and metalloproteases, for use in purification of Histidine-tagged proteins
Expand
View Pricing
P2714
Protease Inhibitor Cocktail,
lyophilized powder, for the inhibition of serine, cysteine and metallo-proteases, for general use, lyophilized powder
Expand
View Pricing
P8849
Protease Inhibitor Cocktail,
DMSO solution, for the inhibition of serine, cysteine, aspartic, aminopeptidases and thermolysin-like activities, for use in purification of Histidine-tagged proteins, DMSO solution
Expand
View Pricing
P9599
Protease Inhibitor Cocktail,
DMSO solution, for the inhibition of serine, cysteine, aspartic, metalloproteases and aminopeptidases, for plant cell and tissue extracts, DMSO solution
Expand
View Pricing
S8820
Protease Inhibitor Cocktail,
SIGMA FAST, tablet, for the inhibition of serine, cysteine, aspartic and metalloproteases, For General Use
Expand
View Pricing
P1860
Protease Inhibitor Cocktail,
DMSO solution, for the inhibition of serine, cysteine, aspartic proteases and aminopeptidases, for use in tissue culture media, DMSO solution
Expand
View Pricing
P8465
Protease Inhibitor Cocktail,
lyophilized powder, for the inhibition of serine, cysteine, aspartic, metalloproteases and aminopeptidases, for use with bacterial cell extracts, lyophilized powder
Expand
View Pricing
P8215
Protease Inhibitor Cocktail,
DMSO solution, for the inhibition of serine, cysteine, aspartic and metalloproteases, for use with fungal and yeast extracts, DMSO solution
Expand
View Pricing
PIC0002
Protease Inhibitor Cocktail,
ReadyShield
®
ready-to-use solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, for use with mammalian cell and tissue extracts, Non-freezing solution
Expand
View Pricing
I3786
Protease Inhibitor Cocktail,
Animal-Free, DMSO solution, for the inhibition of serine, cysteine, aspartic proteases and aminopeptidases, for use with mammalian cell and tissue extract, DMSO solution
Expand
View Pricing
PIC0007
Protease Inhibitor Cocktail,
ReadyShield
®
ready-to-use solution, for the inhibition of serine, cysteine, aspartic and metalloproteases, Safe for Proteomic Mass Spectrometry applications
Expand
View Pricing
I3911
Protease Inhibitor Cocktail,
Animal-Free, lyophilized powder, for the inhibition of serine, cysteine, aspartic and metalloproteases, for general use, lyophilized powder
Expand
View Pricing
PIC0006
Protease Inhibitor Cocktail,
ReadyShield
®
ready-to-use solution, for the inhibition of serine, cysteine, aspartic proteases and aminopeptidases, for use in tissue culture media, Non-freezing solution
Expand
View Pricing
PIC0005
Protease Inhibitor Cocktail,
ReadyShield
®
ready-to-use solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, For plant cell and tissue extracts, Non-freezing solution
Expand
View Pricing
PIC0004
Protease Inhibitor Cocktail,
ReadyShield
®
ready-to-use solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, for use in purification of Histidine-tagged proteins, Non-freezing solution
Expand
View Pricing