Skip to Content
Merck
All Photos(1)

Key Documents

S2075

Sigma-Aldrich

Src Inhibitor-1

≥98% (HPLC)

Synonym(s):

4-(4′-Phenoxyanilino)-6,7-dimethoxyquinazoline, 6,7-Dimethoxy-N-(4-phenoxyphenyl)-4-quinazolinamine, SKI-1, Src-l1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O3
CAS Number:
Molecular Weight:
373.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc2ncnc(Nc3ccc(Oc4ccccc4)cc3)c2cc1OC

InChI

1S/C22H19N3O3/c1-26-20-12-18-19(13-21(20)27-2)23-14-24-22(18)25-15-8-10-17(11-9-15)28-16-6-4-3-5-7-16/h3-14H,1-2H3,(H,23,24,25)

InChI key

DMWVGXGXHPOEPT-UHFFFAOYSA-N

Application

Src Inhibitor-1 may be used in Src kinase-mediated cell signaling studies.

Biochem/physiol Actions

Apart from Src kinase, some Src inhibitors also inhibit the activity of Abl kinase, Lck, Csk and Yes proteins and are therefore being explored as viable anti-cancer drug options.
Src Inhibitor-1 is a potent, competitve dual site (both the ATP- and peptide-binding) Src kinase inhibitor.
Src Inhibitor-1 is a potent, competitve dual site (both the ATP- and peptide-binding) Src kinase inhibitor. Src Inhibitor-1 is one of the "gold standards" for Src kinase inhibition. It has been shown to use PP1 or PP2 in parallel with Src-I1 to inhbit Src family kinases. Src Inhibitor-1 is a selective, competitive inhibitor of Src family of tyrosine kinases (IC50 = 44 nM and 88 nM for Src and Lck, respectively). Src Inhibitor-1 Inhibits VEGFR2 and c-fms tyrosine kinases at much higher concentrations (IC50 = 0.32 μM and 30 μM, respectively).

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diane H Boschelli et al.
Journal of medicinal chemistry, 47(7), 1599-1601 (2004-03-19)
We previously reported that several 7-alkoxy-4-phenylamino-3-quinolinecarbonitriles were potent inhibitors of Src kinase activity. We disclose here a new highly efficient and versatile route to these compounds, which are also potent inhibitors of Abl kinase.
Lei Chen et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 57, 72-83 (2019-01-23)
Periploca forrestii(PF) is mainly utilized for treatment of arthritis and traumatic injury historically. We had previously demonstrated that a fraction rich in cardiotonic steroids isolated from PF had the potential to facilitate wound healing. However, the exact material basis and
Klaske M Schukken et al.
Life science alliance, 3(2) (2020-01-26)
Chromosomal instability (CIN) and aneuploidy are hallmarks of cancer. As most cancers are aneuploid, targeting aneuploidy or CIN may be an effective way to target a broad spectrum of cancers. Here, we perform two small molecule compound screens to identify
Yu Ree Choi et al.
EMBO reports, 21(7), e48950-e48950 (2020-05-07)
Parkinson's disease (PD) is characterized by the loss of dopaminergic neurons located in the substantia nigra pars compacta and the presence of proteinaceous inclusions called Lewy bodies and Lewy neurites in numerous brain regions. Increasing evidence indicates that Lewy pathology
Changli Wei et al.
The Journal of clinical investigation, 129(5), 1946-1959 (2019-02-08)
Soluble urokinase plasminogen activator receptor (suPAR) is an immune-derived circulating signaling molecule that has been implicated in chronic kidney disease, such as focal segmental glomerulosclerosis (FSGS). Typically, native uPAR (isoform 1) translates to a 3-domain protein capable of binding and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service