Skip to Content
Merck
All Photos(1)

Key Documents

SML0837

Sigma-Aldrich

FAK Inhibitor 14

≥95% (HPLC)

Synonym(s):

1,2,4,5-Benzene tetraamine tetrahydrochloride, 1,2,4,5-Benzenetetraamine 4HCl, 1,2,4,5-Benzenetetraamine tetrahydrochloride, Benzene-1,2,4,5-tetramine tetrahydrochloride, NSC 667249, Y15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N4 · 4HCl
CAS Number:
Molecular Weight:
284.01
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

, faint purple to dark brown

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[Cl-].[Cl-].[Cl-].[Cl-].Nc1c(cc(c(c1)N)N)N.[H+].[H+].[H+].[H+]

InChI

1S/C6H10N4.4ClH/c7-3-1-4(8)6(10)2-5(3)9;;;;/h1-2H,7-10H2;4*1H

InChI key

BZDGCIJWPWHAOF-UHFFFAOYSA-N

Application

FAK Inhibitor 14 has been used in the metastasis assay. It is also used to study its effect on bone morphogenetic protein 7 (BMP-7)-induced cell crawling and adhesion.

Biochem/physiol Actions

1,2,4,5-Benzenetetraamine tetrahydrochloride (FAK Inhibitor 14; Y15) is a cell-permeable, selective focal adhesion kinase (FAK) inhibitor.
1,2,4,5-Benzenetetraamine tetrahydrochloride (FAK Inhibitor 14; Y15) is a cell-permeable, selective focal adhesion kinase (FAK) inhibitor. Focal adhesion kinase (FAK) is essential in regulating integrin signaling pathways responsible for cell survival, cell proliferation and motility. Phosphorylation of FAK tyrosine 397 (Y397) forms a high affinity binding site for the SH2 domain of the Src family kinases and PI3 kinase. FAK is overexpressed in a number of human tumors. 1,2,4,5-Benzenetetraamine tetrahydrochloride (FAK Inhibitor 14, Y15) blocks phosphorylation of Y397-FAK, which results in neuroblastoma cell detachment and apoptosis.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Fak page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Sovershaev et al.
Thrombosis research, 147, 24-31 (2016-09-27)
Bone morphogenetic protein (BMP) 7 is abundant in atherosclerotic plaques and increases monocyte pro-coagulant activity by enhancing tissue factor (TF) expression. While several members of the BMP superfamily are able to serve as chemotactic agents for monocytes, the role of
Sophie Bachy et al.
iScience, 25(2), 103758-103758 (2022-02-12)
Macrophages play an important role in immune and matrix regulation during pancreatic adenocarcinoma (PDAC). Collagen deposition massively contributes to the physical and functional changes of the tissue during pathogenesis. We investigated the impact of thick collagen fibers on the phenotype
The adhesion g-Protein-coupled receptor, gPr56/aDgrg1, inhibits cell?extracellular Matrix signaling to Prevent Metastatic Melanoma growth.
Millar M W, et al.
Frontiers in Oncology, 8, 8-8 (2018)
Xiaoling Wang et al.
International journal of molecular sciences, 22(18) (2021-09-29)
By employing an innovative biohybrid membrane, the present study aimed at elucidating the mechanistic role of the focal adhesion kinase (FAK) in epithelial morphogenesis in vitro over 4, 7, and 10 days. The consequences of siRNA-mediated FAK knockdown on epithelial
Erna-Zulaikha Dayang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 133, 111073-111073 (2021-01-01)
Sepsis is a life-threatening condition often leading to multiple organ failure for which currently no pharmacological treatment is available. Endothelial cells (EC) are among the first cells to respond to pathogens and inflammatory mediators in sepsis and might be a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service