Skip to Content
Merck
All Photos(1)

Key Documents

SML1027

Sigma-Aldrich

Imatinib mesylate

≥98% (HPLC), powder, tyrosine kinase inhibitor

Synonym(s):

4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-phenyl]benzamide methanesulfonate, 4-[(4-Methylpiperazin-1-yl)methyl]-N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)benzamide mesylate, CGP 57148, Genfatinib, Glivec, Imatinib methanesulfonate, STI-571

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31N7O · CH4O3S
CAS Number:
Molecular Weight:
589.71
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Imatinib mesylate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])C.[N+H]1(CCN(CC1)Cc2ccc(cc2)C(=O)Nc3cc(c(cc3)C)Nc4nc(ccn4)c5cnccc5)C

InChI

1S/C29H31N7O.CH4O3S/c1-21-5-10-25(18-27(21)34-29-31-13-11-26(33-29)24-4-3-12-30-19-24)32-28(37)23-8-6-22(7-9-23)20-36-16-14-35(2)15-17-36;1-5(2,3)4/h3-13,18-19H,14-17,20H2,1-2H3,(H,32,37)(H,31,33,34);1H3,(H,2,3,4)

InChI key

YLMAHDNUQAMNNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Imatinib mesylate (IM) has been used:
  • in sulforhodamine B cell viability assay to study its effect on CD34+ or CD38- and CD34+ or CD38+ cells
  • as an abelson1 (ABL1) kinase inhibitor
  • in circular dichroism analysis to study the effect of IM binding on the secondary structure of the human transferrins

Biochem/physiol Actions

Imatinib mesylate is a tyrosine kinase inhibitor with antineoplastic activity.
Imatinib mesylate is a tyrosine kinase inhibitor with antineoplastic activity. Imatinib is a potent inhibitor of the Bcr-Abl kinase encoded by the bcr-abl oncogene as well as receptor tyrosine kinases encoded by c-kit and platelet-derived growth factor receptor (PDGFR) oncogenes. Imatinib mesylate inhibition of Bcr-Abl tyrosine kinase created by the Philadelphia chromosome abnormality found in CML decreases proliferation and enhances apoptosis in leukemias CML and ALL. Inhibition of c-kit tyrosine activity inhibits mast-cell and cellular proliferation in those diseases overexpressing c-kit such as gastrointestinal stromal tumor (GIST).

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

Imatinib has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the Imatinib probe summary on the Chemical Probes Portal website.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optogenetic reconstitution reveals that Dynein-Dynactin-NuMA clusters generate cortical spindle-pulling forces as a multi-arm ensemble.
Okumura M, et al.
bioRxiv, 277202-277202 (2018)
Wei Ye et al.
Oncotarget, 8(50), 87002-87015 (2016-07-28)
Available therapeutic options for advanced B cell precursor acute lymphoblastic leukemia (pre-B ALL) are limited. Many lead to neutropenia, leaving patients at risk of life-threatening infections and result in bad outcomes. New treatment options are needed to improve overall survival.
Julien Solinc et al.
Journal of the American Heart Association, 11(7), e023021-e023021 (2022-03-30)
Background Platelet-derived growth factor is a major regulator of the vascular remodeling associated with pulmonary arterial hypertension. We previously showed that protein widely 1 (PW1+) vascular progenitor cells participate in early vessel neomuscularization during experimental pulmonary hypertension (PH) and we
Interaction of imatinib mesylate with human serum transferrin: the comparative spectroscopic studies.
Sliwinska-Hill U, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 173, 468-475 (2017)
Ovatodiolide targets chronic myeloid leukemia stem cells by epigenetically upregulating hsa-miR-155, suppressing the BCR-ABL fusion gene and dysregulating the PI3K/AKT/mTOR pathway
Tu Y X, et al.
Oncotarget, 9(3), 3267-3267 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service