RAB0301
Rat IL-4 ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 0.66-160 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Il4(287287)
Related Categories
General description
The Rat IL-4 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of rat IL-4 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Rat IL-4
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Faten A M Abo-Aziza et al.
Veterinary world, 15(5), 1347-1364 (2022-06-30)
Bone marrow-derived mesenchymal stem cells (BM-MSCs) transplantation and their hepatogenic differentiated cells (HDCs) can be applied for liver injury repair by tissue grafting. Regenerative potentiality in liver cirrhosis models was widely investigated; however, immunomodulation and anti-inflammation in acute hepatitis remain
Jing-Ge Zhang et al.
International immunopharmacology, 20(2), 307-315 (2014-04-08)
Cholecystokinin octapeptide (CCK-8), an immunomodulatory peptide, can promote or suppress the development or function of specific CD4(+) T cell subsets by regulating antigen-presenting cell functions. In the current study, we investigated whether CCK-8 exerts a direct effect on T cells
Christine Hartoonian et al.
Molecular biology reports, 41(9), 5943-5952 (2014-06-29)
Plasmid DNA vaccination is a promising vaccine platform for prevention and treatment of infectious disease. Enhancement of the DNA vaccine potency by co-inoculation of immunoadjuvant has been shown to be an effective strategy. Modulation of dendritic cells and T-cells locomotion
Mohamed El-Behi et al.
European journal of immunology, 44(10), 3003-3014 (2014-07-30)
IL-17-secreting CD8(+) T cells (Tc17 cells) have been implicated in immunity to infections, cancer, and autoimmune diseases. Thus far, studies on Tc17 cells have primarily investigated their development from naïve precursors, while the biology of committed Tc17 cells has been
Marwa Abdallah et al.
Experimental dermatology, 23(10), 742-747 (2014-08-12)
Not only macrophages, T-helper (Th)1 and Th2, but also CD4(+) CD25(high) FoxP3(+) regulatory T cells (T-regs) are involved in immune response to Mycobacterium leprae. We aimed to evaluate serum interleukin (IL)-1β and IL-12p70 (macrophage cytokines), interferon-γ (IFN-γ) (Th1 cytokine), IL-4
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service