Skip to Content
Merck
All Photos(1)

Key Documents

PZ0183

Sigma-Aldrich

ERB-041

≥98% (HPLC)

Synonym(s):

2-(3-Fluoro-4-hydroxyphenyl)-7-vinyl-1,3-benzoxazol-5-ol, Prinaberel, WAY-202041

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10FNO3
CAS Number:
Molecular Weight:
271.24
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥25 mg/mL

storage temp.

room temp

SMILES string

OC(C=C1)=C(F)C=C1C2=NC3=CC(O)=CC(C=C)=C3O2

InChI

1S/C15H10FNO3/c1-2-8-5-10(18)7-12-14(8)20-15(17-12)9-3-4-13(19)11(16)6-9/h2-7,18-19H,1H2

InChI key

MQIMZDXIAHJKQP-UHFFFAOYSA-N

Gene Information

human ... ESR2(2100)

Biochem/physiol Actions

ERB-041 is a potent, selective ERβ receptor agonist (IC50 ERβ: 5.4 nM; > 200-fold selective over ERα)
ERB-041 is a potent, selective estrogen ERβ receptor agonist (IC50 ERβ: 5.4 nM; > 200-fold selective over ERα). ERβ plays a minor role in mediating estrogen action in the uterus, the hypothalamus/pituitary, the skeleton, and other classic estrogen target tissues. However, a clear role for ERβ has been established in the ovary, cardiovascular system, and brain as well as in animal models of inflammation including arthritis, endometriosis, inflammatory bowel disease, and sepsis. There is increasing interest in finding ERβ agonists for potential use in a variety of clinical applications without triggering classic estrogenic side effects.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sreenivasan Paruthiyil et al.
PloS one, 4(7), e6271-e6271 (2009-07-18)
Estrogens produce biological effects by interacting with two estrogen receptors, ERalpha and ERbeta. Drugs that selectively target ERalpha or ERbeta might be safer for conditions that have been traditionally treated with non-selective estrogens. Several synthetic and natural ERbeta-selective compounds have
Iuliia Savchuk et al.
PloS one, 8(8), e71722-e71722 (2013-08-24)
It is well known that estrogens and estrogen-like endocrine disruptors can suppress steroidogenic gene expression, attenuate androgen production and decrease differentiation of adult Leydig cell lineage. However, there is no information about the possible link between the potency of Leydig
Qian Jiang et al.
Purinergic signalling, 13(1), 105-117 (2016-11-07)
Estrogen receptor beta (ERβ) has been shown to play a therapeutic role in inflammatory bowel disease (IBD). However, the mechanism underlying how ERβ exerts therapeutic effects and its relationship with P2X3 receptors (P2X3R) in rats with inflammation is not known.
Geriolda Topi et al.
The Journal of pathology, 251(3), 297-309 (2020-04-26)
Oestrogen receptor β (ERβ) has been suggested to have anti-proliferative and anti-tumour effects in breast and prostate cancer cells, but other studies have indicated its tumour-promoting effects. Understanding the complex effects of this receptor in different contexts requires further study.
Ana Paola G Lombardi et al.
Frontiers in endocrinology, 11, 184-184 (2020-04-25)
Prostate cancer is initially dependent on the androgen, gradually evolves into an androgen-independent form of the disease, also known as castration-resistant prostate cancer (CRPC). At this stage, current therapies scantily improve survival of the patient. Androgens and estrogens are involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service